Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 66801000-66801257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSL5
Entrez Gene ID
10022
Gene Name
insulin like 5
Gene Aliases
PRO182, UNQ156
UniGene
Hs.251380
Gene Chromosome Location
Chr.1: 66797741-66801259 on Build GRCh38
Transcripts
NM_005478.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911763 c.104G>A p.R35Q Chr.1: 66801118-66801118 on Build GRCh38
4756803 c.103C>T p.R35W Chr.1: 66801119-66801119 on Build GRCh38
911764 c.57A>C p.E19D Chr.1: 66801165-66801165 on Build GRCh38
911761 c.143G>A p.R48K Chr.1: 66801079-66801079 on Build GRCh38
911762 c.110T>C p.V37A Chr.1: 66801112-66801112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935972765 Chr.1: 66801202-66801202 on Build GRCh38
rs759887485 Chr.1: 66801182-66801182 on Build GRCh38
rs972884689 Chr.1: 66801184-66801184 on Build GRCh38
rs751867732 Chr.1: 66801127-66801127 on Build GRCh38
rs766779218 Chr.1: 66801111-66801111 on Build GRCh38
rs752782229 Chr.1: 66801033-66801033 on Build GRCh38
rs776128153 Chr.1: 66801203-66801203 on Build GRCh38
rs745534239 Chr.1: 66801147-66801147 on Build GRCh38
rs760065052 Chr.1: 66801114-66801114 on Build GRCh38
rs762133136 Chr.1: 66801219-66801219 on Build GRCh38
rs756036584 Chr.1: 66801047-66801047 on Build GRCh38
rs145962489 Chr.1: 66801122-66801122 on Build GRCh38
rs372170547 Chr.1: 66801064-66801064 on Build GRCh38
rs760971008 Chr.1: 66801122-66801122 on Build GRCh38
rs767795234 Chr.1: 66801226-66801226 on Build GRCh38
rs774042811 Chr.1: 66801120-66801120 on Build GRCh38
rs146406445 Chr.1: 66801161-66801161 on Build GRCh38
rs540801660 Chr.1: 66801103-66801103 on Build GRCh38
rs1013617635 Chr.1: 66801117-66801117 on Build GRCh38
rs376698421 Chr.1: 66801119-66801119 on Build GRCh38
rs746688347 Chr.1: 66801172-66801172 on Build GRCh38
rs763333410 Chr.1: 66801211-66801211 on Build GRCh38
rs764281088 Chr.1: 66801215-66801215 on Build GRCh38
rs771471853 Chr.1: 66801101-66801101 on Build GRCh38
rs777993270 Chr.1: 66801038-66801038 on Build GRCh38
rs770529302 Chr.1: 66801175-66801175 on Build GRCh38
rs747218792 Chr.1: 66801041-66801041 on Build GRCh38
rs903687338 Chr.1: 66801086-66801086 on Build GRCh38
rs139349255 Chr.1: 66801199-66801199 on Build GRCh38
rs749263556 Chr.1: 66801048-66801048 on Build GRCh38
rs867546192 Chr.1: 66801197-66801198 on Build GRCh38
rs752257034 Chr.1: 66801180-66801180 on Build GRCh38
rs982820455 Chr.1: 66801100-66801100 on Build GRCh38
rs778970586 Chr.1: 66801156-66801156 on Build GRCh38
rs144353627 Chr.1: 66801191-66801191 on Build GRCh38
rs149194547 Chr.1: 66801115-66801115 on Build GRCh38
rs137933631 Chr.1: 66801160-66801160 on Build GRCh38
rs141211720 Chr.1: 66801140-66801140 on Build GRCh38
rs760289872 Chr.1: 66801106-66801106 on Build GRCh38
rs754889887 Chr.1: 66801152-66801152 on Build GRCh38
rs776931095 Chr.1: 66801113-66801113 on Build GRCh38
rs765987055 Chr.1: 66801118-66801118 on Build GRCh38
rs748032013 Chr.1: 66801158-66801158 on Build GRCh38
rs758552332 Chr.1: 66801035-66801035 on Build GRCh38
rs549148 Chr.1: 66801073-66801073 on Build GRCh38
rs146856578 Chr.1: 66801141-66801141 on Build GRCh38
rs751653318 Chr.1: 66801218-66801218 on Build GRCh38

View Full Product Details