Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53594160-53594403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS1
Entrez Gene ID
148979
Gene Name
GLIS family zinc finger 1
Gene Aliases
-
UniGene
Hs.306691
Gene Chromosome Location
Chr.1: 53506233-53739171 on Build GRCh38
Transcripts
NM_147193.2 XM_017000408.1 XM_017000409.1 XM_017000410.1 XM_017000411.1 XM_017000412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4988592 c.593G>A p.R198H Chr.1: 53594310-53594310 on Build GRCh38
291390 c.670C>A p.R224S Chr.1: 53594233-53594233 on Build GRCh38
6671611 c.611C>T p.A204V Chr.1: 53594292-53594292 on Build GRCh38
3751299 c.560C>G p.A187G Chr.1: 53594343-53594343 on Build GRCh38
1208306 c.712C>T p.R238C Chr.1: 53594191-53594191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048147750 Chr.1: 53594334-53594334 on Build GRCh38
rs748404280 Chr.1: 53594299-53594299 on Build GRCh38
rs1032416709 Chr.1: 53594213-53594213 on Build GRCh38
rs778828067 Chr.1: 53594368-53594368 on Build GRCh38
rs111547294 Chr.1: 53594376-53594376 on Build GRCh38
rs902458303 Chr.1: 53594375-53594375 on Build GRCh38
rs756693688 Chr.1: 53594255-53594255 on Build GRCh38
rs539985102 Chr.1: 53594239-53594239 on Build GRCh38
rs773798489 Chr.1: 53594227-53594227 on Build GRCh38
rs1007822292 Chr.1: 53594339-53594339 on Build GRCh38
rs569988372 Chr.1: 53594327-53594327 on Build GRCh38
rs193201988 Chr.1: 53594323-53594323 on Build GRCh38
rs375635801 Chr.1: 53594381-53594381 on Build GRCh38
rs57378215 Chr.1: 53594184-53594184 on Build GRCh38
rs774675510 Chr.1: 53594286-53594286 on Build GRCh38
rs370468534 Chr.1: 53594254-53594254 on Build GRCh38
rs761232530 Chr.1: 53594232-53594232 on Build GRCh38
rs749938080 Chr.1: 53594330-53594330 on Build GRCh38
rs749215456 Chr.1: 53594359-53594359 on Build GRCh38
rs766864383 Chr.1: 53594233-53594233 on Build GRCh38
rs370494270 Chr.1: 53594382-53594382 on Build GRCh38
rs997401513 Chr.1: 53594190-53594190 on Build GRCh38
rs747050302 Chr.1: 53594380-53594380 on Build GRCh38
rs200339816 Chr.1: 53594258-53594258 on Build GRCh38
rs35227000 Chr.1: 53594343-53594343 on Build GRCh38
rs766894332 Chr.1: 53594315-53594315 on Build GRCh38
rs878859840 Chr.1: 53594194-53594194 on Build GRCh38
rs149706629 Chr.1: 53594322-53594322 on Build GRCh38
rs917416018 Chr.1: 53594240-53594240 on Build GRCh38
rs754771890 Chr.1: 53594360-53594360 on Build GRCh38
rs1038760577 Chr.1: 53594332-53594332 on Build GRCh38
rs767709665 Chr.1: 53594226-53594226 on Build GRCh38
rs763691093 Chr.1: 53594246-53594246 on Build GRCh38
rs780663060 Chr.1: 53594262-53594262 on Build GRCh38
rs375343014 Chr.1: 53594311-53594311 on Build GRCh38
rs147448341 Chr.1: 53594202-53594202 on Build GRCh38
rs1030668665 Chr.1: 53594195-53594195 on Build GRCh38
rs990922501 Chr.1: 53594228-53594228 on Build GRCh38
rs773361849 Chr.1: 53594379-53594379 on Build GRCh38
rs751276401 Chr.1: 53594373-53594373 on Build GRCh38
rs756875095 Chr.1: 53594326-53594326 on Build GRCh38
rs977945259 Chr.1: 53594208-53594208 on Build GRCh38
rs755350809 Chr.1: 53594242-53594242 on Build GRCh38
rs755594736 Chr.1: 53594338-53594338 on Build GRCh38
rs145375877 Chr.1: 53594277-53594277 on Build GRCh38
rs201038184 Chr.1: 53594283-53594283 on Build GRCh38
rs772471069 Chr.1: 53594310-53594310 on Build GRCh38
rs761944746 Chr.1: 53594224-53594224 on Build GRCh38
rs756076762 Chr.1: 53594274-53594274 on Build GRCh38
rs949422881 Chr.1: 53594270-53594270 on Build GRCh38
rs140770740 Chr.1: 53594188-53594188 on Build GRCh38
rs745412773 Chr.1: 53594263-53594263 on Build GRCh38
rs1048572803 Chr.1: 53594316-53594316 on Build GRCh38

View Full Product Details