Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157037202-157037427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3 XM_011534515.2 XM_017009339.1 XM_017009340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6169838 c.939C>T p.V313V Chr.5: 157037260-157037260 on Build GRCh38
6169837 c.939C>T p.V313V Chr.5: 157037260-157037260 on Build GRCh38
1064874 c.922C>A p.L308I Chr.5: 157037277-157037277 on Build GRCh38
1064875 c.868G>A p.A290T Chr.5: 157037331-157037331 on Build GRCh38
6546862 c.900C>A p.V300V Chr.5: 157037299-157037299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766988802 Chr.5: 157037297-157037297 on Build GRCh38
rs781525427 Chr.5: 157037226-157037226 on Build GRCh38
rs41297583 Chr.5: 157037235-157037235 on Build GRCh38
rs759172493 Chr.5: 157037388-157037388 on Build GRCh38
rs769814175 Chr.5: 157037343-157037343 on Build GRCh38
rs764960630 Chr.5: 157037382-157037383 on Build GRCh38
rs751126401 Chr.5: 157037295-157037295 on Build GRCh38
rs779091220 Chr.5: 157037244-157037244 on Build GRCh38
rs773091016 Chr.5: 157037277-157037277 on Build GRCh38
rs199758684 Chr.5: 157037281-157037281 on Build GRCh38
rs1045555833 Chr.5: 157037373-157037373 on Build GRCh38
rs375393335 Chr.5: 157037299-157037299 on Build GRCh38
rs756566853 Chr.5: 157037302-157037302 on Build GRCh38
rs778055969 Chr.5: 157037234-157037234 on Build GRCh38
rs370221161 Chr.5: 157037250-157037250 on Build GRCh38
rs753176589 Chr.5: 157037227-157037227 on Build GRCh38
rs746027499 Chr.5: 157037327-157037327 on Build GRCh38
rs768697828 Chr.5: 157037257-157037257 on Build GRCh38
rs758620224 Chr.5: 157037349-157037349 on Build GRCh38
rs771103519 Chr.5: 157037238-157037238 on Build GRCh38
rs200398021 Chr.5: 157037333-157037333 on Build GRCh38
rs948631462 Chr.5: 157037356-157037356 on Build GRCh38
rs376460912 Chr.5: 157037309-157037309 on Build GRCh38
rs540137630 Chr.5: 157037393-157037393 on Build GRCh38
rs757526794 Chr.5: 157037312-157037312 on Build GRCh38
rs765913899 Chr.5: 157037288-157037288 on Build GRCh38
rs776426988 Chr.5: 157037262-157037262 on Build GRCh38
rs758926533 Chr.5: 157037369-157037371 on Build GRCh38
rs936981418 Chr.5: 157037361-157037361 on Build GRCh38
rs753161972 Chr.5: 157037298-157037298 on Build GRCh38
rs200556401 Chr.5: 157037296-157037296 on Build GRCh38
rs374244803 Chr.5: 157037377-157037377 on Build GRCh38
rs773883034 Chr.5: 157037381-157037381 on Build GRCh38
rs769722548 Chr.5: 157037268-157037268 on Build GRCh38
rs754171803 Chr.5: 157037311-157037311 on Build GRCh38
rs1054421100 Chr.5: 157037339-157037339 on Build GRCh38
rs749064806 Chr.5: 157037375-157037375 on Build GRCh38
rs761775390 Chr.5: 157037264-157037264 on Build GRCh38
rs2036402 Chr.5: 157037231-157037231 on Build GRCh38
rs370862249 Chr.5: 157037332-157037332 on Build GRCh38
rs778990365 Chr.5: 157037322-157037322 on Build GRCh38
rs922736975 Chr.5: 157037272-157037272 on Build GRCh38

View Full Product Details