Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89060166-89060391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP1
Entrez Gene ID
2633
Gene Name
guanylate binding protein 1
Gene Aliases
-
UniGene
Hs.62661
Gene Chromosome Location
Chr.1: 89052304-89065360 on Build GRCh38
Transcripts
NM_002053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912962 c.294C>T p.T98T Chr.1: 89060221-89060221 on Build GRCh38
5614074 c.208A>G p.T70A Chr.1: 89060307-89060307 on Build GRCh38
3790321 c.199C>A p.L67M Chr.1: 89060316-89060316 on Build GRCh38
5053734 c.270C>T p.G90G Chr.1: 89060245-89060245 on Build GRCh38
1237789 c.251C>T p.P84L Chr.1: 89060264-89060264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369238719 Chr.1: 89060278-89060278 on Build GRCh38
rs536896435 Chr.1: 89060202-89060202 on Build GRCh38
rs770116818 Chr.1: 89060324-89060324 on Build GRCh38
rs1005134972 Chr.1: 89060286-89060286 on Build GRCh38
rs763435415 Chr.1: 89060276-89060276 on Build GRCh38
rs149168400 Chr.1: 89060305-89060305 on Build GRCh38
rs778820979 Chr.1: 89060215-89060215 on Build GRCh38
rs865916158 Chr.1: 89060210-89060210 on Build GRCh38
rs376417084 Chr.1: 89060221-89060221 on Build GRCh38
rs756101749 Chr.1: 89060213-89060213 on Build GRCh38
rs771822601 Chr.1: 89060344-89060344 on Build GRCh38
rs780607200 Chr.1: 89060327-89060327 on Build GRCh38
rs372508569 Chr.1: 89060285-89060285 on Build GRCh38
rs1016473956 Chr.1: 89060301-89060301 on Build GRCh38
rs377536917 Chr.1: 89060347-89060347 on Build GRCh38
rs373495119 Chr.1: 89060340-89060340 on Build GRCh38
rs1048401 Chr.1: 89060283-89060283 on Build GRCh38
rs781557585 Chr.1: 89060229-89060229 on Build GRCh38
rs934541783 Chr.1: 89060236-89060236 on Build GRCh38
rs183022787 Chr.1: 89060349-89060349 on Build GRCh38
rs776570020 Chr.1: 89060353-89060353 on Build GRCh38
rs143075008 Chr.1: 89060220-89060220 on Build GRCh38
rs900562283 Chr.1: 89060270-89060270 on Build GRCh38
rs768227259 Chr.1: 89060333-89060333 on Build GRCh38
rs774025055 Chr.1: 89060335-89060335 on Build GRCh38
rs752796598 Chr.1: 89060216-89060216 on Build GRCh38
rs1012104994 Chr.1: 89060252-89060252 on Build GRCh38
rs764118857 Chr.1: 89060365-89060365 on Build GRCh38
rs746319193 Chr.1: 89060234-89060234 on Build GRCh38
rs759490849 Chr.1: 89060354-89060354 on Build GRCh38
rs757114516 Chr.1: 89060193-89060193 on Build GRCh38
rs146259663 Chr.1: 89060217-89060217 on Build GRCh38
rs777963244 Chr.1: 89060218-89060218 on Build GRCh38
rs186011210 Chr.1: 89060251-89060251 on Build GRCh38
rs751731582 Chr.1: 89060302-89060302 on Build GRCh38
rs746223852 Chr.1: 89060318-89060318 on Build GRCh38
rs138685353 Chr.1: 89060204-89060204 on Build GRCh38
rs142106364 Chr.1: 89060238-89060238 on Build GRCh38
rs138624937 Chr.1: 89060306-89060306 on Build GRCh38
rs141853315 Chr.1: 89060292-89060292 on Build GRCh38
rs878907541 Chr.1: 89060265-89060265 on Build GRCh38
rs753525240 Chr.1: 89060281-89060281 on Build GRCh38
rs749764403 Chr.1: 89060331-89060331 on Build GRCh38
rs374862163 Chr.1: 89060266-89060266 on Build GRCh38
rs762511701 Chr.1: 89060264-89060264 on Build GRCh38
rs774084209 Chr.1: 89060267-89060267 on Build GRCh38
rs191255112 Chr.1: 89060348-89060348 on Build GRCh38
rs751315579 Chr.1: 89060190-89060190 on Build GRCh38
rs543309651 Chr.1: 89060268-89060268 on Build GRCh38
rs767374348 Chr.1: 89060273-89060273 on Build GRCh38

View Full Product Details