Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83429439-83429664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
UniGene
Hs.480101
Gene Chromosome Location
Chr.4: 83407343-83455883 on Build GRCh38
Transcripts
NM_001297755.1 NM_001297756.1 NM_001297757.1 NM_133636.3 XM_005262711.1 XM_005262713.2 XM_006714076.2 XM_017007679.1 XM_017007680.1 XM_017007682.1 XM_017007683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545573 c.2467G>A p.E823K Chr.4: 83429575-83429575 on Build GRCh38
3606607 c.2396C>A p.T799N Chr.4: 83429646-83429646 on Build GRCh38
2955982 c.2461G>A p.E821K Chr.4: 83429581-83429581 on Build GRCh38
7370947 c.2494A>G p.K832E Chr.4: 83429548-83429548 on Build GRCh38
5043006 c.2388G>A p.W796* Chr.4: 83429654-83429654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747408542 Chr.4: 83429491-83429491 on Build GRCh38
rs781742056 Chr.4: 83429634-83429634 on Build GRCh38
rs772624401 Chr.4: 83429483-83429483 on Build GRCh38
rs779841368 Chr.4: 83429597-83429597 on Build GRCh38
rs148996864 Chr.4: 83429539-83429539 on Build GRCh38
rs767524211 Chr.4: 83429585-83429585 on Build GRCh38
rs372487292 Chr.4: 83429625-83429625 on Build GRCh38
rs754683972 Chr.4: 83429605-83429605 on Build GRCh38
rs75238520 Chr.4: 83429586-83429586 on Build GRCh38
rs1049037418 Chr.4: 83429555-83429555 on Build GRCh38
rs778224985 Chr.4: 83429612-83429612 on Build GRCh38
rs753612557 Chr.4: 83429598-83429598 on Build GRCh38
rs776524223 Chr.4: 83429508-83429508 on Build GRCh38
rs200219888 Chr.4: 83429556-83429556 on Build GRCh38
rs1031680651 Chr.4: 83429484-83429484 on Build GRCh38
rs1010903119 Chr.4: 83429505-83429505 on Build GRCh38
rs771300521 Chr.4: 83429633-83429633 on Build GRCh38
rs74836884 Chr.4: 83429501-83429501 on Build GRCh38
rs775735529 Chr.4: 83429548-83429548 on Build GRCh38
rs755735927 Chr.4: 83429589-83429589 on Build GRCh38
rs759691915 Chr.4: 83429538-83429538 on Build GRCh38
rs761611311 Chr.4: 83429581-83429581 on Build GRCh38
rs762719876 Chr.4: 83429550-83429550 on Build GRCh38
rs778421152 Chr.4: 83429488-83429488 on Build GRCh38
rs751369936 Chr.4: 83429573-83429573 on Build GRCh38
rs998191467 Chr.4: 83429629-83429629 on Build GRCh38
rs763940386 Chr.4: 83429569-83429569 on Build GRCh38

View Full Product Details