Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67452614-67452873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP4
Entrez Gene ID
57010
Gene Name
calcium binding protein 4
Gene Aliases
CRSD, CSNB2B
UniGene
Hs.143036
Gene Chromosome Location
Chr.11: 67452406-67461774 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Gene Symbol
GPR152
Entrez Gene ID
390212
Gene Name
G protein-coupled receptor 152
Gene Aliases
PGR5
UniGene
Hs.567997
Gene Chromosome Location
Chr.11: 67451301-67452729 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5894932 c.93G>A p.W31* Chr.11: 67452632-67452632 on Build GRCh38
5894933 c.92G>A p.W31* Chr.11: 67452633-67452633 on Build GRCh38
4903073 c.44G>A p.R15K Chr.11: 67452681-67452681 on Build GRCh38
5747284 c.50G>A p.R17H Chr.11: 67452675-67452675 on Build GRCh38
1298500 c.22G>A p.D8N Chr.11: 67452703-67452703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758304706 Chr.11: 67452751-67452751 on Build GRCh38
rs929793572 Chr.11: 67452666-67452666 on Build GRCh38
rs754851637 Chr.11: 67452721-67452721 on Build GRCh38
rs140961380 Chr.11: 67452712-67452712 on Build GRCh38
rs1038429610 Chr.11: 67452642-67452642 on Build GRCh38
rs757144127 Chr.11: 67452643-67452643 on Build GRCh38
rs963454480 Chr.11: 67452725-67452725 on Build GRCh38
rs201224247 Chr.11: 67452730-67452730 on Build GRCh38
rs769034394 Chr.11: 67452769-67452769 on Build GRCh38
rs377022444 Chr.11: 67452676-67452676 on Build GRCh38
rs550308484 Chr.11: 67452677-67452677 on Build GRCh38
rs550973757 Chr.11: 67452712-67452725 on Build GRCh38
rs180993428 Chr.11: 67452808-67452808 on Build GRCh38
rs746068270 Chr.11: 67452639-67452639 on Build GRCh38
rs749943043 Chr.11: 67452696-67452696 on Build GRCh38
rs960925188 Chr.11: 67452807-67452807 on Build GRCh38
rs200263880 Chr.11: 67452737-67452737 on Build GRCh38
rs367585954 Chr.11: 67452762-67452762 on Build GRCh38
rs373833267 Chr.11: 67452673-67452673 on Build GRCh38
rs374006561 Chr.11: 67452753-67452753 on Build GRCh38
rs763317826 Chr.11: 67452667-67452667 on Build GRCh38
rs370267462 Chr.11: 67452771-67452771 on Build GRCh38
rs769975157 Chr.11: 67452647-67452647 on Build GRCh38
rs189793824 Chr.11: 67452752-67452752 on Build GRCh38
rs150212410 Chr.11: 67452842-67452842 on Build GRCh38
rs574318144 Chr.11: 67452848-67452848 on Build GRCh38
rs373042252 Chr.11: 67452802-67452802 on Build GRCh38
rs1638560 Chr.11: 67452704-67452704 on Build GRCh38
rs199732498 Chr.11: 67452649-67452649 on Build GRCh38
rs774874245 Chr.11: 67452675-67452675 on Build GRCh38
rs556292542 Chr.11: 67452847-67452847 on Build GRCh38
rs781177395 Chr.11: 67452636-67452636 on Build GRCh38
rs925004429 Chr.11: 67452781-67452781 on Build GRCh38
rs201087097 Chr.11: 67452772-67452772 on Build GRCh38
rs897278849 Chr.11: 67452643-67452643 on Build GRCh38
rs936388312 Chr.11: 67452812-67452812 on Build GRCh38
rs370828822 Chr.11: 67452634-67452634 on Build GRCh38
rs749616302 Chr.11: 67452763-67452763 on Build GRCh38
rs760372344 Chr.11: 67452700-67452700 on Build GRCh38

View Full Product Details