Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5906241-5906466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO18
Entrez Gene ID
84893
Gene Name
F-box protein, helicase, 18
Gene Aliases
FBH1, Fbx18, hFBH1
UniGene
Hs.498543
Gene Chromosome Location
Chr.10: 5889572-5937595 on Build GRCh38
Transcripts
NM_001258452.1 NM_001258453.1 NM_032807.4 NM_178150.2 XM_011519748.2 XM_017016842.1 XM_017016843.1 XM_017016844.1 XM_017016845.1 XM_017016846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4500422 c.564C>T p.T188T Chr.10: 5906290-5906290 on Build GRCh38
6886030 c.577G>A p.G193R Chr.10: 5906303-5906303 on Build GRCh38
5699574 c.698A>G p.Q233R Chr.10: 5906424-5906424 on Build GRCh38
5979839 c.712G>C p.V238L Chr.10: 5906438-5906438 on Build GRCh38
340880 c.572G>T p.W191L Chr.10: 5906298-5906298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937670689 Chr.10: 5906378-5906378 on Build GRCh38
rs755192280 Chr.10: 5906324-5906324 on Build GRCh38
rs745428782 Chr.10: 5906348-5906348 on Build GRCh38
rs781058434 Chr.10: 5906424-5906424 on Build GRCh38
rs779335043 Chr.10: 5906326-5906326 on Build GRCh38
rs777546393 Chr.10: 5906339-5906339 on Build GRCh38
rs145958035 Chr.10: 5906394-5906394 on Build GRCh38
rs143001244 Chr.10: 5906393-5906393 on Build GRCh38
rs956119911 Chr.10: 5906432-5906432 on Build GRCh38
rs201826348 Chr.10: 5906420-5906420 on Build GRCh38
rs755673158 Chr.10: 5906437-5906437 on Build GRCh38
rs761110537 Chr.10: 5906316-5906316 on Build GRCh38
rs139867605 Chr.10: 5906404-5906404 on Build GRCh38
rs143135958 Chr.10: 5906411-5906411 on Build GRCh38
rs146670441 Chr.10: 5906431-5906431 on Build GRCh38
rs753866866 Chr.10: 5906320-5906320 on Build GRCh38
rs766440649 Chr.10: 5906317-5906317 on Build GRCh38
rs767981499 Chr.10: 5906302-5906302 on Build GRCh38
rs762146468 Chr.10: 5906382-5906382 on Build GRCh38
rs201891050 Chr.10: 5906419-5906419 on Build GRCh38
rs752531900 Chr.10: 5906330-5906330 on Build GRCh38
rs368256849 Chr.10: 5906376-5906376 on Build GRCh38
rs773482471 Chr.10: 5906388-5906388 on Build GRCh38
rs748533038 Chr.10: 5906285-5906285 on Build GRCh38
rs769432989 Chr.10: 5906354-5906354 on Build GRCh38
rs578126156 Chr.10: 5906289-5906289 on Build GRCh38
rs763885556 Chr.10: 5906413-5906413 on Build GRCh38
rs750988878 Chr.10: 5906304-5906304 on Build GRCh38
rs138692839 Chr.10: 5906389-5906389 on Build GRCh38
rs770780965 Chr.10: 5906341-5906341 on Build GRCh38
rs780697016 Chr.10: 5906345-5906345 on Build GRCh38
rs758304318 Chr.10: 5906337-5906337 on Build GRCh38
rs141566367 Chr.10: 5906284-5906284 on Build GRCh38
rs150913018 Chr.10: 5906294-5906294 on Build GRCh38
rs545497071 Chr.10: 5906300-5906300 on Build GRCh38
rs759164198 Chr.10: 5906288-5906288 on Build GRCh38
rs753016172 Chr.10: 5906409-5906409 on Build GRCh38
rs746993373 Chr.10: 5906369-5906369 on Build GRCh38
rs373590165 Chr.10: 5906262-5906262 on Build GRCh38
rs768195671 Chr.10: 5906441-5906441 on Build GRCh38
rs796644562 Chr.10: 5906259-5906259 on Build GRCh38
rs772381502 Chr.10: 5906383-5906383 on Build GRCh38
rs887914766 Chr.10: 5906397-5906397 on Build GRCh38
rs759756063 Chr.10: 5906403-5906403 on Build GRCh38
rs148412195 Chr.10: 5906438-5906438 on Build GRCh38
rs1056166310 Chr.10: 5906381-5906381 on Build GRCh38

View Full Product Details