Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92463760-92464031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683610 c.2675G>A p.R892H Chr.10: 92463817-92463817 on Build GRCh38
6055348 c.2585G>A p.R862K Chr.10: 92463907-92463907 on Build GRCh38
3441529 c.2496C>T p.I832I Chr.10: 92463996-92463996 on Build GRCh38
372117 c.2648A>T p.Q883L Chr.10: 92463844-92463844 on Build GRCh38
3441528 c.2610G>T p.M870I Chr.10: 92463882-92463882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036877168 Chr.10: 92463815-92463815 on Build GRCh38
rs756421508 Chr.10: 92463824-92463824 on Build GRCh38
rs774700889 Chr.10: 92463847-92463847 on Build GRCh38
rs199882899 Chr.10: 92463977-92463977 on Build GRCh38
rs1051901293 Chr.10: 92463809-92463809 on Build GRCh38
rs747216756 Chr.10: 92463871-92463871 on Build GRCh38
rs762604985 Chr.10: 92463813-92463813 on Build GRCh38
rs61736443 Chr.10: 92463969-92463969 on Build GRCh38
rs759157349 Chr.10: 92463957-92463957 on Build GRCh38
rs764585694 Chr.10: 92463968-92463968 on Build GRCh38
rs1012143879 Chr.10: 92463893-92463893 on Build GRCh38
rs747306678 Chr.10: 92464006-92464006 on Build GRCh38
rs761870451 Chr.10: 92463899-92463899 on Build GRCh38
rs766038121 Chr.10: 92463923-92463923 on Build GRCh38
rs995278812 Chr.10: 92463849-92463849 on Build GRCh38
rs772780300 Chr.10: 92463915-92463915 on Build GRCh38
rs192230461 Chr.10: 92463889-92463889 on Build GRCh38
rs1042444 Chr.10: 92463959-92463959 on Build GRCh38
rs547079065 Chr.10: 92463922-92463922 on Build GRCh38
rs780254461 Chr.10: 92463998-92463998 on Build GRCh38
rs757789861 Chr.10: 92463982-92463982 on Build GRCh38
rs779064748 Chr.10: 92463852-92463852 on Build GRCh38
rs768540270 Chr.10: 92463883-92463883 on Build GRCh38
rs750552917 Chr.10: 92463995-92463995 on Build GRCh38
rs769557547 Chr.10: 92463909-92463909 on Build GRCh38
rs751261195 Chr.10: 92463953-92463953 on Build GRCh38
rs200287247 Chr.10: 92463867-92463867 on Build GRCh38
rs753087364 Chr.10: 92463821-92463821 on Build GRCh38
rs374667709 Chr.10: 92463861-92463861 on Build GRCh38
rs758600182 Chr.10: 92463996-92463996 on Build GRCh38
rs749885722 Chr.10: 92463792-92463792 on Build GRCh38
rs764872364 Chr.10: 92463789-92463789 on Build GRCh38
rs535825881 Chr.10: 92463987-92463987 on Build GRCh38
rs940536511 Chr.10: 92463980-92463980 on Build GRCh38
rs984686097 Chr.10: 92463944-92463944 on Build GRCh38
rs757697787 Chr.10: 92463851-92463851 on Build GRCh38
rs765946014 Chr.10: 92463814-92463814 on Build GRCh38
rs150078715 Chr.10: 92463846-92463846 on Build GRCh38
rs145451690 Chr.10: 92463860-92463860 on Build GRCh38
rs776288772 Chr.10: 92463894-92463894 on Build GRCh38
rs761559283 Chr.10: 92463941-92463941 on Build GRCh38

View Full Product Details