Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91055932-91056193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
UniGene
Hs.125750
Gene Chromosome Location
Chr.12: 91050491-91058354 on Build GRCh38
Transcripts
NM_007035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940282 c.91G>A p.D31N Chr.12: 91056191-91056191 on Build GRCh38
1364825 c.327C>T p.N109N Chr.12: 91055955-91055955 on Build GRCh38
7373403 c.286C>A p.Q96K Chr.12: 91055996-91055996 on Build GRCh38
4453208 c.265A>C p.K89Q Chr.12: 91056017-91056017 on Build GRCh38
7299767 c.305T>C p.L102P Chr.12: 91055977-91055977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564854065 Chr.12: 91056026-91056026 on Build GRCh38
rs761217146 Chr.12: 91056041-91056041 on Build GRCh38
rs770668689 Chr.12: 91056023-91056023 on Build GRCh38
rs779203403 Chr.12: 91056061-91056061 on Build GRCh38
rs917190458 Chr.12: 91056012-91056012 on Build GRCh38
rs567010026 Chr.12: 91056067-91056067 on Build GRCh38
rs2735334 Chr.12: 91056009-91056009 on Build GRCh38
rs766988573 Chr.12: 91056036-91056036 on Build GRCh38
rs769750761 Chr.12: 91056001-91056001 on Build GRCh38
rs377426809 Chr.12: 91056039-91056039 on Build GRCh38
rs527407560 Chr.12: 91056057-91056057 on Build GRCh38
rs745496599 Chr.12: 91056102-91056102 on Build GRCh38
rs778007693 Chr.12: 91056071-91056071 on Build GRCh38
rs755097045 Chr.12: 91056081-91056081 on Build GRCh38
rs772179610 Chr.12: 91055990-91055990 on Build GRCh38
rs756325200 Chr.12: 91055996-91055996 on Build GRCh38
rs991300088 Chr.12: 91056002-91056002 on Build GRCh38
rs376196047 Chr.12: 91055987-91055987 on Build GRCh38
rs903143563 Chr.12: 91056110-91056110 on Build GRCh38
rs372534447 Chr.12: 91056011-91056011 on Build GRCh38
rs751660260 Chr.12: 91056100-91056100 on Build GRCh38
rs35693771 Chr.12: 91056075-91056075 on Build GRCh38
rs199813838 Chr.12: 91056162-91056162 on Build GRCh38
rs754279432 Chr.12: 91055958-91055958 on Build GRCh38
rs376135902 Chr.12: 91056060-91056060 on Build GRCh38
rs2540003 Chr.12: 91056018-91056018 on Build GRCh38
rs775332060 Chr.12: 91056111-91056111 on Build GRCh38
rs760233605 Chr.12: 91056113-91056113 on Build GRCh38
rs758552587 Chr.12: 91056073-91056073 on Build GRCh38
rs547054836 Chr.12: 91056058-91056058 on Build GRCh38
rs777653993 Chr.12: 91056083-91056083 on Build GRCh38
rs777263244 Chr.12: 91056121-91056121 on Build GRCh38
rs765570180 Chr.12: 91056122-91056122 on Build GRCh38
rs771645894 Chr.12: 91056106-91056106 on Build GRCh38
rs1041498553 Chr.12: 91056079-91056079 on Build GRCh38
rs757611751 Chr.12: 91055962-91055962 on Build GRCh38
rs770704095 Chr.12: 91056099-91056099 on Build GRCh38
rs762711254 Chr.12: 91056022-91056022 on Build GRCh38
rs2735335 Chr.12: 91055973-91055973 on Build GRCh38
rs202094304 Chr.12: 91056154-91056154 on Build GRCh38
rs751817390 Chr.12: 91056161-91056161 on Build GRCh38
rs779165739 Chr.12: 91055967-91055967 on Build GRCh38
rs954905600 Chr.12: 91055979-91055979 on Build GRCh38
rs748011083 Chr.12: 91055997-91055997 on Build GRCh38
rs867739680 Chr.12: 91056136-91056136 on Build GRCh38
rs750783253 Chr.12: 91056129-91056129 on Build GRCh38
rs938609998 Chr.12: 91056144-91056144 on Build GRCh38
rs544690510 Chr.12: 91055996-91055996 on Build GRCh38
rs746142009 Chr.12: 91055976-91055976 on Build GRCh38
rs138807366 Chr.12: 91056077-91056077 on Build GRCh38
rs758594193 Chr.12: 91056151-91056151 on Build GRCh38
rs149529940 Chr.12: 91056027-91056027 on Build GRCh38
rs868610169 Chr.12: 91055994-91055994 on Build GRCh38
rs764675474 Chr.12: 91056050-91056050 on Build GRCh38
rs763765245 Chr.12: 91056115-91056115 on Build GRCh38

View Full Product Details