Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44302941-44303186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS2
Entrez Gene ID
57505
Gene Name
alanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
AARSL, COXPD8, LKENP, MT-ALARS, MTALARS
UniGene
Hs.158381
Gene Chromosome Location
Chr.6: 44297850-44313358 on Build GRCh38
Transcripts
NM_020745.3 XM_005249245.3 XM_011514764.2 XM_017011112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
484142 c.2255+1G>T p.? Chr.6: 44303065-44303065 on Build GRCh38
6880117 c.2181G>A p.G727G Chr.6: 44303140-44303140 on Build GRCh38
6281125 c.2151C>A p.Y717* Chr.6: 44303170-44303170 on Build GRCh38
4999552 c.2188G>A p.V730M Chr.6: 44303133-44303133 on Build GRCh38
7267005 c.2254A>G p.T752A Chr.6: 44303067-44303067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899381542 Chr.6: 44302976-44302976 on Build GRCh38
rs762226722 Chr.6: 44303162-44303162 on Build GRCh38
rs372806295 Chr.6: 44303116-44303116 on Build GRCh38
rs951180412 Chr.6: 44303011-44303011 on Build GRCh38
rs777436220 Chr.6: 44303146-44303146 on Build GRCh38
rs863223861 Chr.6: 44303114-44303114 on Build GRCh38
rs325002 Chr.6: 44303031-44303031 on Build GRCh38
rs1014113579 Chr.6: 44302983-44302983 on Build GRCh38
rs750729510 Chr.6: 44303085-44303085 on Build GRCh38
rs775065697 Chr.6: 44303159-44303159 on Build GRCh38
rs147163018 Chr.6: 44303101-44303101 on Build GRCh38
rs778875250 Chr.6: 44303126-44303126 on Build GRCh38
rs546686376 Chr.6: 44303026-44303026 on Build GRCh38
rs759449104 Chr.6: 44303019-44303019 on Build GRCh38
rs755023355 Chr.6: 44303103-44303103 on Build GRCh38
rs955887304 Chr.6: 44303092-44303092 on Build GRCh38
rs765232662 Chr.6: 44303021-44303041 on Build GRCh38
rs890110022 Chr.6: 44303011-44303011 on Build GRCh38
rs78397386 Chr.6: 44303068-44303068 on Build GRCh38
rs776390375 Chr.6: 44303154-44303154 on Build GRCh38
rs759069299 Chr.6: 44303061-44303061 on Build GRCh38
rs79389653 Chr.6: 44303027-44303027 on Build GRCh38
rs752088510 Chr.6: 44303072-44303072 on Build GRCh38
rs1037797275 Chr.6: 44302972-44302972 on Build GRCh38
rs886061486 Chr.6: 44303119-44303119 on Build GRCh38
rs867480864 Chr.6: 44303128-44303128 on Build GRCh38
rs536579476 Chr.6: 44303100-44303100 on Build GRCh38
rs560761240 Chr.6: 44302996-44302996 on Build GRCh38
rs751129588 Chr.6: 44303074-44303074 on Build GRCh38
rs541995956 Chr.6: 44302995-44302995 on Build GRCh38
rs566667883 Chr.6: 44303156-44303156 on Build GRCh38
rs946886065 Chr.6: 44303127-44303127 on Build GRCh38
rs530359122 Chr.6: 44302971-44302971 on Build GRCh38
rs144224923 Chr.6: 44303134-44303134 on Build GRCh38
rs369747415 Chr.6: 44303090-44303090 on Build GRCh38
rs776295527 Chr.6: 44303054-44303054 on Build GRCh38
rs145436818 Chr.6: 44303157-44303157 on Build GRCh38
rs325003 Chr.6: 44302961-44302961 on Build GRCh38
rs370007720 Chr.6: 44303073-44303073 on Build GRCh38
rs770550880 Chr.6: 44303037-44303037 on Build GRCh38
rs1025505743 Chr.6: 44302989-44302989 on Build GRCh38
rs1032837001 Chr.6: 44303091-44303091 on Build GRCh38
rs746761805 Chr.6: 44303153-44303153 on Build GRCh38
rs988173583 Chr.6: 44303108-44303108 on Build GRCh38
rs753952552 Chr.6: 44303102-44303102 on Build GRCh38
rs993704976 Chr.6: 44302991-44302991 on Build GRCh38
rs35623954 Chr.6: 44303133-44303133 on Build GRCh38
rs528141636 Chr.6: 44303017-44303017 on Build GRCh38

View Full Product Details