Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 64224266-64224539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB12
Entrez Gene ID
142689
Gene Name
ankyrin repeat and SOCS box containing 12
Gene Aliases
-
UniGene
Hs.56281
Gene Chromosome Location
Chr.X: 64224192-64230631 on Build GRCh38
Transcripts
NM_130388.3

SNP Detail

SNP ID Chromosome Location
rs201762866 Chr.X: 64224382-64224382 on Build GRCh38
rs986675172 Chr.X: 64224333-64224333 on Build GRCh38
rs779578237 Chr.X: 64224344-64224344 on Build GRCh38
rs748815216 Chr.X: 64224430-64224430 on Build GRCh38
rs148589525 Chr.X: 64224460-64224460 on Build GRCh38
rs759017776 Chr.X: 64224330-64224330 on Build GRCh38
rs778673287 Chr.X: 64224481-64224481 on Build GRCh38
rs1021737414 Chr.X: 64224509-64224509 on Build GRCh38
rs891089634 Chr.X: 64224419-64224419 on Build GRCh38
rs753978819 Chr.X: 64224294-64224303 on Build GRCh38
rs747992783 Chr.X: 64224488-64224488 on Build GRCh38
rs201958583 Chr.X: 64224357-64224357 on Build GRCh38
rs199530232 Chr.X: 64224305-64224305 on Build GRCh38
rs200651920 Chr.X: 64224384-64224384 on Build GRCh38
rs891977541 Chr.X: 64224469-64224469 on Build GRCh38
rs755735038 Chr.X: 64224343-64224343 on Build GRCh38
rs747069332 Chr.X: 64224400-64224400 on Build GRCh38
rs200654794 Chr.X: 64224429-64224429 on Build GRCh38
rs965182812 Chr.X: 64224518-64224518 on Build GRCh38
rs746075982 Chr.X: 64224507-64224507 on Build GRCh38
rs1003576016 Chr.X: 64224439-64224439 on Build GRCh38
rs957821927 Chr.X: 64224295-64224295 on Build GRCh38
rs1055923183 Chr.X: 64224386-64224386 on Build GRCh38
rs753409435 Chr.X: 64224467-64224467 on Build GRCh38
rs1041062156 Chr.X: 64224485-64224485 on Build GRCh38
rs747066776 Chr.X: 64224499-64224499 on Build GRCh38
rs767133351 Chr.X: 64224336-64224336 on Build GRCh38
rs773264171 Chr.X: 64224392-64224392 on Build GRCh38
rs933931567 Chr.X: 64224327-64224327 on Build GRCh38
rs367633622 Chr.X: 64224310-64224310 on Build GRCh38
rs144858124 Chr.X: 64224438-64224438 on Build GRCh38
rs759820562 Chr.X: 64224301-64224301 on Build GRCh38
rs140412046 Chr.X: 64224502-64224502 on Build GRCh38
rs931461263 Chr.X: 64224399-64224399 on Build GRCh38
rs1032168263 Chr.X: 64224285-64224285 on Build GRCh38
rs764713991 Chr.X: 64224436-64224436 on Build GRCh38
rs755611319 Chr.X: 64224443-64224443 on Build GRCh38
rs748907381 Chr.X: 64224384-64224384 on Build GRCh38
rs750896461 Chr.X: 64224324-64224324 on Build GRCh38
rs754982702 Chr.X: 64224300-64224300 on Build GRCh38
rs776745282 Chr.X: 64224421-64224421 on Build GRCh38
rs964755793 Chr.X: 64224287-64224287 on Build GRCh38
rs765670083 Chr.X: 64224427-64224427 on Build GRCh38
rs779425447 Chr.X: 64224303-64224303 on Build GRCh38
rs759856795 Chr.X: 64224426-64224426 on Build GRCh38
rs149424932 Chr.X: 64224391-64224391 on Build GRCh38
rs749961504 Chr.X: 64224340-64224340 on Build GRCh38
rs758271149 Chr.X: 64224489-64224489 on Build GRCh38
rs771114865 Chr.X: 64224416-64224416 on Build GRCh38
rs778035840 Chr.X: 64224368-64224368 on Build GRCh38
rs753237124 Chr.X: 64224317-64224317 on Build GRCh38
rs139928365 Chr.X: 64224365-64224365 on Build GRCh38
rs777573069 Chr.X: 64224498-64224498 on Build GRCh38

View Full Product Details