Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45137355-45137604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001135705.2 NM_001135706.2 NM_001135707.2 NM_001321352.1 NM_001321353.1 NM_024722.3 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025094.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181562 c.416-2A>G p.? Chr.17: 45137366-45137366 on Build GRCh38
1181561 c.416-2A>G p.? Chr.17: 45137366-45137366 on Build GRCh38
4774487 c.471C>T p.C157C Chr.17: 45137423-45137423 on Build GRCh38
4658771 c.490delC p.P165fs*9 Chr.17: 45137442-45137442 on Build GRCh38
6271729 c.416-5G>T p.? Chr.17: 45137363-45137363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903565882 Chr.17: 45137494-45137494 on Build GRCh38
rs997827078 Chr.17: 45137582-45137582 on Build GRCh38
rs757998128 Chr.17: 45137459-45137459 on Build GRCh38
rs377135057 Chr.17: 45137437-45137437 on Build GRCh38
rs555285235 Chr.17: 45137529-45137529 on Build GRCh38
rs902831303 Chr.17: 45137378-45137378 on Build GRCh38
rs766415785 Chr.17: 45137426-45137426 on Build GRCh38
rs372896376 Chr.17: 45137424-45137424 on Build GRCh38
rs779170657 Chr.17: 45137497-45137497 on Build GRCh38
rs373038659 Chr.17: 45137382-45137382 on Build GRCh38
rs755333763 Chr.17: 45137496-45137496 on Build GRCh38
rs749837964 Chr.17: 45137484-45137484 on Build GRCh38
rs774604967 Chr.17: 45137402-45137402 on Build GRCh38
rs764971732 Chr.17: 45137450-45137450 on Build GRCh38
rs751078601 Chr.17: 45137396-45137398 on Build GRCh38
rs1028186515 Chr.17: 45137522-45137522 on Build GRCh38
rs891949786 Chr.17: 45137471-45137471 on Build GRCh38
rs768955668 Chr.17: 45137398-45137398 on Build GRCh38
rs1017660765 Chr.17: 45137580-45137580 on Build GRCh38
rs755978060 Chr.17: 45137482-45137482 on Build GRCh38
rs748352757 Chr.17: 45137404-45137404 on Build GRCh38
rs375050946 Chr.17: 45137581-45137581 on Build GRCh38
rs752344621 Chr.17: 45137454-45137454 on Build GRCh38
rs772144831 Chr.17: 45137406-45137406 on Build GRCh38
rs756815371 Chr.17: 45137479-45137479 on Build GRCh38
rs748347665 Chr.17: 45137502-45137502 on Build GRCh38
rs759344417 Chr.17: 45137445-45137445 on Build GRCh38
rs763590435 Chr.17: 45137475-45137475 on Build GRCh38
rs773440975 Chr.17: 45137423-45137423 on Build GRCh38
rs780511863 Chr.17: 45137481-45137481 on Build GRCh38
rs756615556 Chr.17: 45137376-45137376 on Build GRCh38
rs780601484 Chr.17: 45137383-45137383 on Build GRCh38
rs759181471 Chr.17: 45137442-45137442 on Build GRCh38
rs751062514 Chr.17: 45137477-45137477 on Build GRCh38
rs998486782 Chr.17: 45137427-45137427 on Build GRCh38
rs375678654 Chr.17: 45137396-45137396 on Build GRCh38
rs772189553 Chr.17: 45137504-45137504 on Build GRCh38
rs779979781 Chr.17: 45137540-45137540 on Build GRCh38
rs1032685494 Chr.17: 45137428-45137428 on Build GRCh38
rs955453432 Chr.17: 45137542-45137542 on Build GRCh38
rs1007244865 Chr.17: 45137500-45137500 on Build GRCh38

View Full Product Details