Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44290808-44291033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
UniGene
Hs.129829
Gene Chromosome Location
Chr.21: 44285838-44298219 on Build GRCh38
Transcripts
NM_000383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6597286 c.50G>T p.R17I Chr.21: 44290856-44290856 on Build GRCh38
6342609 c.125G>T p.G42V Chr.21: 44290931-44290931 on Build GRCh38
6597287 c.22G>A p.G8R Chr.21: 44290828-44290828 on Build GRCh38
1592790 c.158G>A p.G53E Chr.21: 44290964-44290964 on Build GRCh38
5449515 c.59T>G p.V20G Chr.21: 44290865-44290865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766896493 Chr.21: 44291011-44291011 on Build GRCh38
rs768040995 Chr.21: 44291000-44291000 on Build GRCh38
rs752100711 Chr.21: 44290879-44290879 on Build GRCh38
rs764403208 Chr.21: 44290884-44290884 on Build GRCh38
rs770013170 Chr.21: 44290912-44290921 on Build GRCh38
rs1001352930 Chr.21: 44290869-44290869 on Build GRCh38
rs145284511 Chr.21: 44290861-44290861 on Build GRCh38
rs200437331 Chr.21: 44290836-44290836 on Build GRCh38
rs1024048649 Chr.21: 44290852-44290852 on Build GRCh38
rs773057776 Chr.21: 44290842-44290842 on Build GRCh38
rs758103414 Chr.21: 44290922-44290922 on Build GRCh38
rs139012063 Chr.21: 44290921-44290921 on Build GRCh38
rs781020126 Chr.21: 44290956-44290956 on Build GRCh38
rs1054395568 Chr.21: 44290977-44290977 on Build GRCh38
rs543275794 Chr.21: 44290828-44290828 on Build GRCh38
rs750913835 Chr.21: 44290892-44290892 on Build GRCh38
rs1016946107 Chr.21: 44290971-44290971 on Build GRCh38
rs369035241 Chr.21: 44290832-44290832 on Build GRCh38
rs934466755 Chr.21: 44290942-44290942 on Build GRCh38
rs1034129728 Chr.21: 44290875-44290875 on Build GRCh38
rs762268160 Chr.21: 44290887-44290887 on Build GRCh38
rs745835455 Chr.21: 44290960-44290960 on Build GRCh38
rs763154847 Chr.21: 44290882-44290882 on Build GRCh38
rs954417560 Chr.21: 44290877-44290877 on Build GRCh38
rs565153757 Chr.21: 44290979-44290979 on Build GRCh38
rs750768166 Chr.21: 44291009-44291009 on Build GRCh38
rs769768095 Chr.21: 44290962-44290962 on Build GRCh38
rs747802771 Chr.21: 44290835-44290835 on Build GRCh38
rs369810277 Chr.21: 44290944-44290944 on Build GRCh38
rs765283449 Chr.21: 44290872-44290872 on Build GRCh38
rs776581217 Chr.21: 44290862-44290862 on Build GRCh38
rs781592011 Chr.21: 44290890-44290890 on Build GRCh38
rs142297552 Chr.21: 44291010-44291010 on Build GRCh38
rs140965390 Chr.21: 44290897-44290897 on Build GRCh38
rs763110689 Chr.21: 44290967-44290967 on Build GRCh38
rs961517445 Chr.21: 44290858-44290858 on Build GRCh38
rs775611325 Chr.21: 44290878-44290878 on Build GRCh38
rs777407927 Chr.21: 44290938-44290938 on Build GRCh38
rs778324245 Chr.21: 44290900-44290900 on Build GRCh38
rs762075832 Chr.21: 44290990-44290990 on Build GRCh38
rs770737578 Chr.21: 44290955-44290955 on Build GRCh38
rs866470102 Chr.21: 44290895-44290895 on Build GRCh38
rs751778017 Chr.21: 44290886-44290886 on Build GRCh38
rs775495071 Chr.21: 44290965-44290965 on Build GRCh38
rs754240163 Chr.21: 44291012-44291012 on Build GRCh38
rs547235267 Chr.21: 44290890-44290890 on Build GRCh38
rs137910738 Chr.21: 44290985-44290985 on Build GRCh38
rs757773290 Chr.21: 44290889-44290890 on Build GRCh38
rs771680046 Chr.21: 44290841-44290841 on Build GRCh38

View Full Product Details