Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38716901-38717158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
UniGene
Hs.270291
Gene Chromosome Location
Chr.19: 38647616-38730532 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3533550 c.941C>T p.P314L Chr.19: 38717114-38717114 on Build GRCh38
7352481 c.929G>A p.R310Q Chr.19: 38717102-38717102 on Build GRCh38
6328247 c.975C>T p.I325I Chr.19: 38717148-38717148 on Build GRCh38
1481023 c.979G>A p.E327K Chr.19: 38717152-38717152 on Build GRCh38
4512772 c.915C>T p.L305L Chr.19: 38717088-38717088 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs146936990 Chr.19: 38717121-38717121 on Build GRCh38
rs771187908 Chr.19: 38717035-38717035 on Build GRCh38
rs1048393899 Chr.19: 38716933-38716933 on Build GRCh38
rs949925069 Chr.19: 38716957-38716957 on Build GRCh38
rs569030532 Chr.19: 38717039-38717039 on Build GRCh38
rs879024343 Chr.19: 38716998-38716998 on Build GRCh38
rs267605466 Chr.19: 38717112-38717112 on Build GRCh38
rs774148634 Chr.19: 38717053-38717053 on Build GRCh38
rs112545413 Chr.19: 38717102-38717102 on Build GRCh38
rs555942368 Chr.19: 38716964-38716970 on Build GRCh38
rs750154056 Chr.19: 38716964-38716964 on Build GRCh38
rs368471159 Chr.19: 38717003-38717003 on Build GRCh38
rs778625571 Chr.19: 38717109-38717109 on Build GRCh38
rs948196744 Chr.19: 38716970-38716970 on Build GRCh38
rs1003352558 Chr.19: 38717022-38717022 on Build GRCh38
rs185752145 Chr.19: 38716942-38716942 on Build GRCh38
rs764202475 Chr.19: 38717051-38717051 on Build GRCh38
rs557293843 Chr.19: 38717012-38717012 on Build GRCh38
rs546315191 Chr.19: 38717064-38717064 on Build GRCh38
rs893885967 Chr.19: 38717058-38717058 on Build GRCh38
rs1002621300 Chr.19: 38717025-38717025 on Build GRCh38
rs756003995 Chr.19: 38717101-38717101 on Build GRCh38
rs746223131 Chr.19: 38717042-38717042 on Build GRCh38
rs796992678 Chr.19: 38717019-38717020 on Build GRCh38
rs267605467 Chr.19: 38717113-38717113 on Build GRCh38
rs567679429 Chr.19: 38716934-38716934 on Build GRCh38
rs879283421 Chr.19: 38716936-38716936 on Build GRCh38
rs754506208 Chr.19: 38717106-38717106 on Build GRCh38
rs559700598 Chr.19: 38717105-38717105 on Build GRCh38
rs750162681 Chr.19: 38717065-38717065 on Build GRCh38
rs763002280 Chr.19: 38717047-38717047 on Build GRCh38
rs376113967 Chr.19: 38717046-38717046 on Build GRCh38
rs761866160 Chr.19: 38717063-38717063 on Build GRCh38
rs757716490 Chr.19: 38717115-38717115 on Build GRCh38
rs1047580489 Chr.19: 38716939-38716939 on Build GRCh38

View Full Product Details