Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141655446-141655707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP3
Entrez Gene ID
64411
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Gene Aliases
CENTD3, DRAG1
UniGene
Hs.726187
Gene Chromosome Location
Chr.5: 141653401-141682233 on Build GRCh38
Transcripts
NM_022481.5 XM_005268497.1 XM_005268498.1 XM_005268499.1 XM_006714792.1 XM_011537677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3409954 c.4025C>T p.A1342V Chr.5: 141655706-141655706 on Build GRCh38
1310787 c.4094C>T p.S1365F Chr.5: 141655637-141655637 on Build GRCh38
4851661 c.4030C>T p.Q1344* Chr.5: 141655701-141655701 on Build GRCh38
5772914 c.4110+10C>G p.? Chr.5: 141655611-141655611 on Build GRCh38
1063531 c.4058C>A p.P1353H Chr.5: 141655673-141655673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775700507 Chr.5: 141655577-141655577 on Build GRCh38
rs746250184 Chr.5: 141655614-141655614 on Build GRCh38
rs972458607 Chr.5: 141655580-141655580 on Build GRCh38
rs148152980 Chr.5: 141655639-141655639 on Build GRCh38
rs142211119 Chr.5: 141655671-141655671 on Build GRCh38
rs758768178 Chr.5: 141655658-141655658 on Build GRCh38
rs540037842 Chr.5: 141655541-141655541 on Build GRCh38
rs2306340 Chr.5: 141655681-141655681 on Build GRCh38
rs771830541 Chr.5: 141655576-141655576 on Build GRCh38
rs375543507 Chr.5: 141655593-141655593 on Build GRCh38
rs114921133 Chr.5: 141655583-141655583 on Build GRCh38
rs376503540 Chr.5: 141655514-141655514 on Build GRCh38
rs766718819 Chr.5: 141655605-141655605 on Build GRCh38
rs768856824 Chr.5: 141655655-141655655 on Build GRCh38
rs529225343 Chr.5: 141655589-141655589 on Build GRCh38
rs753385527 Chr.5: 141655621-141655621 on Build GRCh38
rs750878671 Chr.5: 141655600-141655600 on Build GRCh38
rs768750584 Chr.5: 141655586-141655586 on Build GRCh38
rs1034442017 Chr.5: 141655473-141655473 on Build GRCh38
rs755797274 Chr.5: 141655608-141655608 on Build GRCh38
rs774834869 Chr.5: 141655686-141655686 on Build GRCh38
rs138955685 Chr.5: 141655663-141655663 on Build GRCh38
rs145534243 Chr.5: 141655631-141655631 on Build GRCh38
rs1021915849 Chr.5: 141655563-141655563 on Build GRCh38
rs776656679 Chr.5: 141655588-141655588 on Build GRCh38
rs377203403 Chr.5: 141655524-141655524 on Build GRCh38
rs55977140 Chr.5: 141655682-141655682 on Build GRCh38
rs201780469 Chr.5: 141655667-141655667 on Build GRCh38
rs79464052 Chr.5: 141655607-141655607 on Build GRCh38
rs147712909 Chr.5: 141655649-141655649 on Build GRCh38
rs1010897051 Chr.5: 141655498-141655498 on Build GRCh38
rs763746634 Chr.5: 141655610-141655610 on Build GRCh38
rs529010789 Chr.5: 141655584-141655584 on Build GRCh38
rs778718658 Chr.5: 141655634-141655634 on Build GRCh38
rs758147299 Chr.5: 141655642-141655642 on Build GRCh38
rs1035650779 Chr.5: 141655481-141655481 on Build GRCh38
rs141039995 Chr.5: 141655646-141655646 on Build GRCh38
rs986518079 Chr.5: 141655633-141655633 on Build GRCh38
rs763242488 Chr.5: 141655604-141655604 on Build GRCh38
rs369505822 Chr.5: 141655668-141655668 on Build GRCh38
rs368935621 Chr.5: 141655594-141655594 on Build GRCh38
rs758929341 Chr.5: 141655594-141655609 on Build GRCh38

View Full Product Details