Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10306229-10306490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM
Entrez Gene ID
133
Gene Name
adrenomedullin
Gene Aliases
AM, PAMP
UniGene
Hs.441047
Gene Chromosome Location
Chr.11: 10304980-10307402 on Build GRCh38
Transcripts
NM_001124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687261 c.310G>T p.G104C Chr.11: 10306393-10306393 on Build GRCh38
3808199 c.261C>G p.A87A Chr.11: 10306344-10306344 on Build GRCh38
7132239 c.249-8G>C p.? Chr.11: 10306324-10306324 on Build GRCh38
3791116 c.338G>A p.G113E Chr.11: 10306421-10306421 on Build GRCh38
6349866 c.348G>C p.T116T Chr.11: 10306431-10306431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774600873 Chr.11: 10306322-10306322 on Build GRCh38
rs764380862 Chr.11: 10306325-10306325 on Build GRCh38
rs769040940 Chr.11: 10306312-10306318 on Build GRCh38
rs758193916 Chr.11: 10306284-10306284 on Build GRCh38
rs1021128489 Chr.11: 10306336-10306336 on Build GRCh38
rs201286035 Chr.11: 10306327-10306327 on Build GRCh38
rs997886079 Chr.11: 10306419-10306419 on Build GRCh38
rs754185495 Chr.11: 10306279-10306279 on Build GRCh38
rs746111731 Chr.11: 10306362-10306362 on Build GRCh38
rs140844268 Chr.11: 10306468-10306468 on Build GRCh38
rs769338742 Chr.11: 10306374-10306374 on Build GRCh38
rs777662561 Chr.11: 10306347-10306347 on Build GRCh38
rs764564742 Chr.11: 10306274-10306274 on Build GRCh38
rs904900719 Chr.11: 10306251-10306251 on Build GRCh38
rs766043791 Chr.11: 10306289-10306289 on Build GRCh38
rs780346402 Chr.11: 10306371-10306371 on Build GRCh38
rs767155083 Chr.11: 10306441-10306441 on Build GRCh38
rs774351156 Chr.11: 10306255-10306255 on Build GRCh38
rs762219274 Chr.11: 10306333-10306333 on Build GRCh38
rs762191630 Chr.11: 10306400-10306400 on Build GRCh38
rs759638767 Chr.11: 10306263-10306263 on Build GRCh38
rs373413227 Chr.11: 10306349-10306349 on Build GRCh38
rs1004066005 Chr.11: 10306376-10306376 on Build GRCh38
rs751083990 Chr.11: 10306291-10306291 on Build GRCh38
rs962018945 Chr.11: 10306378-10306378 on Build GRCh38
rs866552090 Chr.11: 10306397-10306397 on Build GRCh38
rs780528887 Chr.11: 10306297-10306297 on Build GRCh38
rs748417465 Chr.11: 10306328-10306342 on Build GRCh38
rs777551708 Chr.11: 10306314-10306314 on Build GRCh38
rs764691516 Chr.11: 10306328-10306328 on Build GRCh38
rs754484775 Chr.11: 10306293-10306293 on Build GRCh38
rs778520098 Chr.11: 10306350-10306350 on Build GRCh38
rs2228573 Chr.11: 10306337-10306337 on Build GRCh38
rs774785781 Chr.11: 10306343-10306343 on Build GRCh38
rs752251431 Chr.11: 10306445-10306445 on Build GRCh38
rs776168196 Chr.11: 10306267-10306267 on Build GRCh38
rs771106210 Chr.11: 10306317-10306317 on Build GRCh38
rs751259666 Chr.11: 10306341-10306341 on Build GRCh38
rs112526486 Chr.11: 10306461-10306461 on Build GRCh38
rs747075720 Chr.11: 10306373-10306373 on Build GRCh38
rs1029162865 Chr.11: 10306295-10306295 on Build GRCh38
rs772115980 Chr.11: 10306365-10306365 on Build GRCh38
rs777195912 Chr.11: 10306330-10306330 on Build GRCh38
rs746037060 Chr.11: 10306324-10306324 on Build GRCh38
rs752136972 Chr.11: 10306345-10306345 on Build GRCh38
rs749067732 Chr.11: 10306315-10306315 on Build GRCh38
rs1015392795 Chr.11: 10306383-10306383 on Build GRCh38
rs367972567 Chr.11: 10306431-10306431 on Build GRCh38
rs770217561 Chr.11: 10306415-10306415 on Build GRCh38
rs767028428 Chr.11: 10306344-10306344 on Build GRCh38
rs779061396 Chr.11: 10306317-10306318 on Build GRCh38
rs376677864 Chr.11: 10306375-10306375 on Build GRCh38
rs763554599 Chr.11: 10306466-10306466 on Build GRCh38
rs777250833 Chr.11: 10306379-10306379 on Build GRCh38
rs761211066 Chr.11: 10306269-10306269 on Build GRCh38
rs748107126 Chr.11: 10306298-10306298 on Build GRCh38
rs755988517 Chr.11: 10306312-10306312 on Build GRCh38
rs934960667 Chr.11: 10306256-10306256 on Build GRCh38

View Full Product Details