Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1453047-1453276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6405256 c.102C>A p.S34R Chr.19: 1453103-1453103 on Build GRCh38
6365610 c.131C>T p.S44L Chr.19: 1453132-1453132 on Build GRCh38
709794 c.145G>A p.V49I Chr.19: 1453250-1453250 on Build GRCh38
5646720 c.164G>C p.G55A Chr.19: 1453269-1453269 on Build GRCh38
7088099 c.84G>A p.E28E Chr.19: 1453085-1453085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033810573 Chr.19: 1453188-1453188 on Build GRCh38
rs757989814 Chr.19: 1453178-1453178 on Build GRCh38
rs769872545 Chr.19: 1453130-1453130 on Build GRCh38
rs768209025 Chr.19: 1453151-1453151 on Build GRCh38
rs773529688 Chr.19: 1453132-1453132 on Build GRCh38
rs750054600 Chr.19: 1453172-1453172 on Build GRCh38
rs115348986 Chr.19: 1453164-1453164 on Build GRCh38
rs760110907 Chr.19: 1453149-1453149 on Build GRCh38
rs528977199 Chr.19: 1453133-1453133 on Build GRCh38
rs761943470 Chr.19: 1453116-1453116 on Build GRCh38
rs893880734 Chr.19: 1453080-1453080 on Build GRCh38
rs764735997 Chr.19: 1453226-1453226 on Build GRCh38
rs771120331 Chr.19: 1453203-1453203 on Build GRCh38
rs367639985 Chr.19: 1453237-1453237 on Build GRCh38
rs768116741 Chr.19: 1453214-1453214 on Build GRCh38
rs111830742 Chr.19: 1453191-1453191 on Build GRCh38
rs753367526 Chr.19: 1453158-1453158 on Build GRCh38
rs905470339 Chr.19: 1453152-1453152 on Build GRCh38
rs760023075 Chr.19: 1453207-1453207 on Build GRCh38
rs201231128 Chr.19: 1453073-1453073 on Build GRCh38
rs910531013 Chr.19: 1453230-1453230 on Build GRCh38
rs565521837 Chr.19: 1453160-1453160 on Build GRCh38
rs73516808 Chr.19: 1453161-1453161 on Build GRCh38
rs374526792 Chr.19: 1453225-1453225 on Build GRCh38
rs774855479 Chr.19: 1453206-1453206 on Build GRCh38
rs377491377 Chr.19: 1453194-1453194 on Build GRCh38
rs569977037 Chr.19: 1453179-1453179 on Build GRCh38
rs746864222 Chr.19: 1453182-1453182 on Build GRCh38
rs765282564 Chr.19: 1453187-1453188 on Build GRCh38
rs776679183 Chr.19: 1453094-1453094 on Build GRCh38
rs944625934 Chr.19: 1453241-1453241 on Build GRCh38
rs752710101 Chr.19: 1453220-1453231 on Build GRCh38
rs139475598 Chr.19: 1453135-1453135 on Build GRCh38
rs144976480 Chr.19: 1453224-1453224 on Build GRCh38
rs749912722 Chr.19: 1453227-1453227 on Build GRCh38
rs375278949 Chr.19: 1453193-1453193 on Build GRCh38
rs758055533 Chr.19: 1453236-1453236 on Build GRCh38
rs774871929 Chr.19: 1453147-1453147 on Build GRCh38
rs1052153630 Chr.19: 1453097-1453097 on Build GRCh38

View Full Product Details