Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69153348-69153599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4439215 c.4076C>T p.S1359L Chr.17: 69153365-69153365 on Build GRCh38
5343007 c.4072C>T p.P1358S Chr.17: 69153369-69153369 on Build GRCh38
6589434 c.4070A>G p.N1357S Chr.17: 69153371-69153371 on Build GRCh38
7294477 c.3990G>A p.Q1330Q Chr.17: 69153522-69153522 on Build GRCh38
365104 c.4022C>T p.S1341L Chr.17: 69153490-69153490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758783431 Chr.17: 69153412-69153412 on Build GRCh38
rs371378007 Chr.17: 69153424-69153424 on Build GRCh38
rs753462878 Chr.17: 69153461-69153461 on Build GRCh38
rs761243208 Chr.17: 69153465-69153465 on Build GRCh38
rs768440283 Chr.17: 69153509-69153509 on Build GRCh38
rs750958061 Chr.17: 69153471-69153471 on Build GRCh38
rs766862947 Chr.17: 69153528-69153528 on Build GRCh38
rs199521991 Chr.17: 69153406-69153406 on Build GRCh38
rs764752677 Chr.17: 69153468-69153468 on Build GRCh38
rs367947150 Chr.17: 69153420-69153420 on Build GRCh38
rs201554524 Chr.17: 69153391-69153391 on Build GRCh38
rs369780103 Chr.17: 69153495-69153495 on Build GRCh38
rs201197046 Chr.17: 69153372-69153372 on Build GRCh38
rs368825986 Chr.17: 69153472-69153472 on Build GRCh38
rs772122971 Chr.17: 69153500-69153500 on Build GRCh38
rs372290702 Chr.17: 69153522-69153522 on Build GRCh38
rs12601012 Chr.17: 69153416-69153416 on Build GRCh38
rs755799243 Chr.17: 69153387-69153387 on Build GRCh38
rs938654051 Chr.17: 69153385-69153385 on Build GRCh38
rs138751693 Chr.17: 69153492-69153492 on Build GRCh38
rs753276170 Chr.17: 69153397-69153397 on Build GRCh38
rs1038590696 Chr.17: 69153519-69153519 on Build GRCh38
rs763649386 Chr.17: 69153459-69153459 on Build GRCh38
rs781583604 Chr.17: 69153560-69153560 on Build GRCh38
rs915554439 Chr.17: 69153561-69153561 on Build GRCh38
rs755324546 Chr.17: 69153542-69153542 on Build GRCh38
rs752050480 Chr.17: 69153536-69153536 on Build GRCh38
rs745852400 Chr.17: 69153498-69153498 on Build GRCh38
rs761485868 Chr.17: 69153512-69153512 on Build GRCh38
rs145841711 Chr.17: 69153399-69153399 on Build GRCh38
rs746314562 Chr.17: 69153411-69153411 on Build GRCh38
rs960218649 Chr.17: 69153555-69153555 on Build GRCh38
rs777970498 Chr.17: 69153497-69153497 on Build GRCh38
rs146959347 Chr.17: 69153525-69153525 on Build GRCh38
rs372750572 Chr.17: 69153475-69153475 on Build GRCh38
rs774454802 Chr.17: 69153428-69153428 on Build GRCh38
rs375730362 Chr.17: 69153484-69153484 on Build GRCh38
rs142796433 Chr.17: 69153506-69153506 on Build GRCh38
rs769798989 Chr.17: 69153496-69153496 on Build GRCh38
rs752431471 Chr.17: 69153373-69153373 on Build GRCh38
rs764764140 Chr.17: 69153518-69153518 on Build GRCh38
rs114802155 Chr.17: 69153435-69153435 on Build GRCh38
rs906931014 Chr.17: 69153378-69153378 on Build GRCh38
rs573989632 Chr.17: 69153421-69153421 on Build GRCh38
rs775405557 Chr.17: 69153442-69153442 on Build GRCh38
rs374716314 Chr.17: 69153455-69153455 on Build GRCh38
rs201564034 Chr.17: 69153415-69153415 on Build GRCh38
rs748185220 Chr.17: 69153423-69153423 on Build GRCh38

View Full Product Details