Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19409148-19409373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER2
Entrez Gene ID
340485
Gene Name
alkaline ceramidase 2
Gene Aliases
ALKCDase2, ASAH3L
UniGene
Hs.41379
Gene Chromosome Location
Chr.9: 19408927-19455173 on Build GRCh38
Transcripts
NM_001010887.2 XM_005251448.3 XM_011517858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395732 c.108+8G>T p.? Chr.9: 19409200-19409200 on Build GRCh38
5782557 c.108G>A p.T36T Chr.9: 19409192-19409192 on Build GRCh38
6266658 c.108+1G>T p.? Chr.9: 19409193-19409193 on Build GRCh38
5507629 c.99C>G p.F33L Chr.9: 19409183-19409183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748615867 Chr.9: 19409188-19409188 on Build GRCh38
rs755282852 Chr.9: 19409170-19409170 on Build GRCh38
rs745662574 Chr.9: 19409209-19409209 on Build GRCh38
rs895612226 Chr.9: 19409273-19409273 on Build GRCh38
rs747105650 Chr.9: 19409199-19409199 on Build GRCh38
rs559426338 Chr.9: 19409287-19409287 on Build GRCh38
rs536580498 Chr.9: 19409329-19409329 on Build GRCh38
rs11792865 Chr.9: 19409208-19409208 on Build GRCh38
rs1028618246 Chr.9: 19409330-19409330 on Build GRCh38
rs769611020 Chr.9: 19409178-19409182 on Build GRCh38
rs894471694 Chr.9: 19409243-19409243 on Build GRCh38
rs539250751 Chr.9: 19409349-19409349 on Build GRCh38
rs547822581 Chr.9: 19409306-19409306 on Build GRCh38
rs779397326 Chr.9: 19409185-19409185 on Build GRCh38
rs529654370 Chr.9: 19409296-19409296 on Build GRCh38
rs565095485 Chr.9: 19409242-19409242 on Build GRCh38
rs893364809 Chr.9: 19409323-19409323 on Build GRCh38
rs540752079 Chr.9: 19409276-19409276 on Build GRCh38
rs999006468 Chr.9: 19409291-19409291 on Build GRCh38
rs769668275 Chr.9: 19409210-19409210 on Build GRCh38
rs1026985680 Chr.9: 19409304-19409304 on Build GRCh38
rs1042710824 Chr.9: 19409169-19409169 on Build GRCh38
rs375063645 Chr.9: 19409351-19409351 on Build GRCh38
rs567574615 Chr.9: 19409314-19409314 on Build GRCh38
rs1012719322 Chr.9: 19409281-19409281 on Build GRCh38
rs201611502 Chr.9: 19409201-19409201 on Build GRCh38
rs757755326 Chr.9: 19409248-19409248 on Build GRCh38
rs975566696 Chr.9: 19409334-19409334 on Build GRCh38
rs921455063 Chr.9: 19409335-19409335 on Build GRCh38
rs543593006 Chr.9: 19409221-19409221 on Build GRCh38
rs566355155 Chr.9: 19409328-19409328 on Build GRCh38
rs758496444 Chr.9: 19409191-19409191 on Build GRCh38
rs576417429 Chr.9: 19409212-19409212 on Build GRCh38
rs777766936 Chr.9: 19409192-19409192 on Build GRCh38
rs532197555 Chr.9: 19409247-19409247 on Build GRCh38
rs1046227167 Chr.9: 19409286-19409286 on Build GRCh38
rs147387981 Chr.9: 19409332-19409332 on Build GRCh38
rs571734328 Chr.9: 19409347-19409347 on Build GRCh38
rs1033198121 Chr.9: 19409300-19409300 on Build GRCh38

View Full Product Details