Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25388709-25388950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
33080 c.463A>G p.I155V Chr.4: 25388736-25388736 on Build GRCh38
6570953 c.458C>G p.S153* Chr.4: 25388731-25388731 on Build GRCh38
273965 c.463A>C p.I155L Chr.4: 25388736-25388736 on Build GRCh38
1054667 c.481T>C p.S161P Chr.4: 25388848-25388848 on Build GRCh38
291206 c.462A>G p.K154K Chr.4: 25388735-25388735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369201112 Chr.4: 25388893-25388893 on Build GRCh38
rs772425146 Chr.4: 25388747-25388747 on Build GRCh38
rs140263991 Chr.4: 25388877-25388877 on Build GRCh38
rs753808827 Chr.4: 25388868-25388868 on Build GRCh38
rs759612137 Chr.4: 25388777-25388777 on Build GRCh38
rs764355801 Chr.4: 25388823-25388823 on Build GRCh38
rs755955531 Chr.4: 25388841-25388841 on Build GRCh38
rs373231076 Chr.4: 25388898-25388898 on Build GRCh38
rs375520498 Chr.4: 25388829-25388829 on Build GRCh38
rs778301350 Chr.4: 25388878-25388878 on Build GRCh38
rs1039344227 Chr.4: 25388750-25388750 on Build GRCh38
rs576432227 Chr.4: 25388753-25388753 on Build GRCh38
rs777326490 Chr.4: 25388774-25388774 on Build GRCh38
rs769851498 Chr.4: 25388922-25388922 on Build GRCh38
rs779920658 Chr.4: 25388843-25388843 on Build GRCh38
rs758560430 Chr.4: 25388815-25388815 on Build GRCh38
rs759960480 Chr.4: 25388919-25388919 on Build GRCh38
rs752894560 Chr.4: 25388800-25388800 on Build GRCh38
rs765259045 Chr.4: 25388791-25388791 on Build GRCh38
rs747353957 Chr.4: 25388899-25388899 on Build GRCh38
rs777236882 Chr.4: 25388908-25388908 on Build GRCh38
rs761186712 Chr.4: 25388752-25388752 on Build GRCh38
rs773700623 Chr.4: 25388751-25388751 on Build GRCh38
rs750190478 Chr.4: 25388825-25388825 on Build GRCh38
rs1008728942 Chr.4: 25388861-25388861 on Build GRCh38
rs766961269 Chr.4: 25388754-25388754 on Build GRCh38

View Full Product Details