Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113388916-113389155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAD
Entrez Gene ID
210
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADH, PBGS
UniGene
Hs.1227
Gene Chromosome Location
Chr.9: 113386312-113401338 on Build GRCh38
Transcripts
NM_000031.5 NM_001003945.2 NM_001317745.1 XM_011518364.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422476 c.823G>A p.V275M Chr.9: 113389085-113389085 on Build GRCh38
6422475 c.910G>A p.V304M Chr.9: 113389085-113389085 on Build GRCh38
3382401 c.1009C>G p.R337G Chr.9: 113388986-113388986 on Build GRCh38
3664023 c.822C>T p.A274A Chr.9: 113389086-113389086 on Build GRCh38
3699351 c.897C>T p.A299A Chr.9: 113389011-113389011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751107367 Chr.9: 113389089-113389089 on Build GRCh38
rs757028029 Chr.9: 113389071-113389071 on Build GRCh38
rs373715261 Chr.9: 113389031-113389031 on Build GRCh38
rs150649205 Chr.9: 113389032-113389032 on Build GRCh38
rs367823672 Chr.9: 113389045-113389045 on Build GRCh38
rs913213690 Chr.9: 113388995-113388995 on Build GRCh38
rs556307080 Chr.9: 113388986-113388986 on Build GRCh38
rs371639473 Chr.9: 113389086-113389086 on Build GRCh38
rs750264863 Chr.9: 113389077-113389077 on Build GRCh38
rs200713896 Chr.9: 113389133-113389133 on Build GRCh38
rs745845598 Chr.9: 113389010-113389010 on Build GRCh38
rs778812183 Chr.9: 113389007-113389007 on Build GRCh38
rs539712107 Chr.9: 113388973-113388973 on Build GRCh38
rs202188395 Chr.9: 113389064-113389064 on Build GRCh38
rs370242589 Chr.9: 113389011-113389011 on Build GRCh38
rs201859063 Chr.9: 113389021-113389021 on Build GRCh38
rs769596841 Chr.9: 113389102-113389102 on Build GRCh38
rs372316464 Chr.9: 113388957-113388957 on Build GRCh38
rs36087664 Chr.9: 113388997-113388997 on Build GRCh38
rs775372297 Chr.9: 113389053-113389053 on Build GRCh38
rs764298726 Chr.9: 113389060-113389060 on Build GRCh38
rs200781693 Chr.9: 113389034-113389034 on Build GRCh38
rs771543639 Chr.9: 113388936-113388936 on Build GRCh38
rs893250215 Chr.9: 113388971-113388971 on Build GRCh38
rs759917153 Chr.9: 113389038-113389038 on Build GRCh38
rs749655574 Chr.9: 113389033-113389033 on Build GRCh38
rs773664007 Chr.9: 113388945-113388945 on Build GRCh38
rs886063364 Chr.9: 113388962-113388962 on Build GRCh38
rs993268256 Chr.9: 113388994-113388994 on Build GRCh38
rs905106024 Chr.9: 113389001-113389001 on Build GRCh38
rs775185719 Chr.9: 113388937-113388937 on Build GRCh38
rs746681065 Chr.9: 113388940-113388940 on Build GRCh38
rs757113916 Chr.9: 113388999-113388999 on Build GRCh38
rs779094672 Chr.9: 113389104-113389104 on Build GRCh38
rs200378521 Chr.9: 113389035-113389035 on Build GRCh38
rs1054563732 Chr.9: 113388962-113388962 on Build GRCh38
rs121912981 Chr.9: 113389085-113389085 on Build GRCh38
rs144785407 Chr.9: 113389076-113389076 on Build GRCh38
rs4986829 Chr.9: 113388960-113388960 on Build GRCh38
rs747842090 Chr.9: 113389100-113389100 on Build GRCh38
rs550332573 Chr.9: 113388956-113388956 on Build GRCh38
rs61729493 Chr.9: 113389055-113389055 on Build GRCh38
rs779391805 Chr.9: 113389019-113389019 on Build GRCh38
rs754592118 Chr.9: 113389094-113389094 on Build GRCh38
rs553974879 Chr.9: 113389068-113389068 on Build GRCh38
rs780968225 Chr.9: 113389099-113389099 on Build GRCh38
rs8177817 Chr.9: 113388943-113388943 on Build GRCh38
rs121912983 Chr.9: 113389088-113389088 on Build GRCh38
rs764311002 Chr.9: 113388989-113388989 on Build GRCh38
rs763412262 Chr.9: 113388946-113388946 on Build GRCh38
rs201745784 Chr.9: 113389109-113389109 on Build GRCh38
rs767530480 Chr.9: 113388963-113388963 on Build GRCh38
rs546001979 Chr.9: 113388949-113388949 on Build GRCh38
rs772389560 Chr.9: 113389124-113389124 on Build GRCh38

View Full Product Details