Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34876621-34876866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS2
Entrez Gene ID
55902
Gene Name
acyl-CoA synthetase short-chain family member 2
Gene Aliases
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1
UniGene
Hs.517034
Gene Chromosome Location
Chr.20: 34874942-34927966 on Build GRCh38
Transcripts
NM_001076552.2 NM_001242393.1 NM_018677.3 XM_005260455.1 XM_005260456.1 XM_006723826.1 XM_011528905.1 XM_011528906.1 XM_011528907.1 XM_011528908.1 XM_011528909.2 XM_011528911.1 XM_011528912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5657165 c.129G>T p.S43S Chr.20: 34876774-34876774 on Build GRCh38
4793822 c.52G>A p.E18K Chr.20: 34876697-34876697 on Build GRCh38
395178 c.135G>T p.Q45H Chr.20: 34876780-34876780 on Build GRCh38
5439332 c.31G>A p.G11S Chr.20: 34876676-34876676 on Build GRCh38
4793823 c.52G>A p.E18K Chr.20: 34876697-34876697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779630921 Chr.20: 34876645-34876645 on Build GRCh38
rs940632186 Chr.20: 34876746-34876746 on Build GRCh38
rs965269016 Chr.20: 34876754-34876754 on Build GRCh38
rs764139474 Chr.20: 34876800-34876800 on Build GRCh38
rs778472257 Chr.20: 34876699-34876699 on Build GRCh38
rs758995393 Chr.20: 34876665-34876665 on Build GRCh38
rs769838087 Chr.20: 34876741-34876741 on Build GRCh38
rs1036639738 Chr.20: 34876807-34876807 on Build GRCh38
rs895525657 Chr.20: 34876808-34876808 on Build GRCh38
rs746111837 Chr.20: 34876740-34876740 on Build GRCh38
rs965750662 Chr.20: 34876687-34876687 on Build GRCh38
rs748766067 Chr.20: 34876664-34876664 on Build GRCh38
rs866522476 Chr.20: 34876674-34876674 on Build GRCh38
rs921087686 Chr.20: 34876765-34876765 on Build GRCh38
rs1032387407 Chr.20: 34876706-34876706 on Build GRCh38
rs146486104 Chr.20: 34876769-34876769 on Build GRCh38
rs747486877 Chr.20: 34876725-34876725 on Build GRCh38
rs909607996 Chr.20: 34876787-34876787 on Build GRCh38
rs974483217 Chr.20: 34876695-34876695 on Build GRCh38
rs775580200 Chr.20: 34876757-34876757 on Build GRCh38
rs868794357 Chr.20: 34876652-34876652 on Build GRCh38
rs776841902 Chr.20: 34876732-34876732 on Build GRCh38
rs984380604 Chr.20: 34876779-34876779 on Build GRCh38
rs771388973 Chr.20: 34876731-34876731 on Build GRCh38
rs866610613 Chr.20: 34876649-34876649 on Build GRCh38
rs949660998 Chr.20: 34876833-34876833 on Build GRCh38
rs976733074 Chr.20: 34876755-34876755 on Build GRCh38
rs921573601 Chr.20: 34876703-34876703 on Build GRCh38
rs141085032 Chr.20: 34876842-34876842 on Build GRCh38
rs1002793627 Chr.20: 34876691-34876691 on Build GRCh38

View Full Product Details