Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120254600-120254839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255A
Entrez Gene ID
55026
Gene Name
transmembrane protein 255A
Gene Aliases
FAM70A
UniGene
Hs.437563
Gene Chromosome Location
Chr.X: 120250984-120311556 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

Gene Symbol
ZBTB33
Entrez Gene ID
10009
Gene Name
zinc finger and BTB domain containing 33
Gene Aliases
ZNF-kaiso, ZNF348
UniGene
Hs.143604
Gene Chromosome Location
Chr.X: 120250752-120258396 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

SNP Detail

SNP ID Chromosome Location
rs782191030 Chr.X: 120254793-120254793 on Build GRCh38
rs782272539 Chr.X: 120254813-120254813 on Build GRCh38
rs782705448 Chr.X: 120254731-120254731 on Build GRCh38
rs782463731 Chr.X: 120254728-120254728 on Build GRCh38
rs781859968 Chr.X: 120254800-120254800 on Build GRCh38
rs782477054 Chr.X: 120254786-120254786 on Build GRCh38
rs781904744 Chr.X: 120254736-120254736 on Build GRCh38
rs782537395 Chr.X: 120254741-120254741 on Build GRCh38
rs782121762 Chr.X: 120254649-120254649 on Build GRCh38
rs1032127603 Chr.X: 120254757-120254757 on Build GRCh38
rs782029088 Chr.X: 120254812-120254812 on Build GRCh38
rs781824519 Chr.X: 120254699-120254699 on Build GRCh38
rs782309844 Chr.X: 120254805-120254805 on Build GRCh38
rs782385890 Chr.X: 120254817-120254817 on Build GRCh38
rs782643714 Chr.X: 120254746-120254746 on Build GRCh38
rs267606333 Chr.X: 120254698-120254698 on Build GRCh38
rs782605427 Chr.X: 120254789-120254789 on Build GRCh38
rs782012980 Chr.X: 120254644-120254644 on Build GRCh38
rs115462363 Chr.X: 120254634-120254634 on Build GRCh38
rs372655083 Chr.X: 120254771-120254771 on Build GRCh38

View Full Product Details