Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102870047-102870272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AS3MT
Entrez Gene ID
57412
Gene Name
arsenite methyltransferase
Gene Aliases
CYT19
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102869453-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433810 c.66G>T p.K22N Chr.10: 102870107-102870107 on Build GRCh38
6455079 c.156A>T p.E52D Chr.10: 102870197-102870197 on Build GRCh38
3433811 c.133G>T p.E45* Chr.10: 102870174-102870174 on Build GRCh38
7289475 c.52G>A p.G18R Chr.10: 102870093-102870093 on Build GRCh38
3709815 c.68G>T p.R23I Chr.10: 102870109-102870109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027298145 Chr.10: 102870204-102870204 on Build GRCh38
rs773980641 Chr.10: 102870078-102870078 on Build GRCh38
rs760664254 Chr.10: 102870131-102870131 on Build GRCh38
rs771537950 Chr.10: 102870211-102870211 on Build GRCh38
rs775888270 Chr.10: 102870098-102870098 on Build GRCh38
rs750267547 Chr.10: 102870128-102870128 on Build GRCh38
rs377652199 Chr.10: 102870243-102870243 on Build GRCh38
rs763309223 Chr.10: 102870108-102870109 on Build GRCh38
rs1020351796 Chr.10: 102870149-102870149 on Build GRCh38
rs779145933 Chr.10: 102870178-102870178 on Build GRCh38
rs369401059 Chr.10: 102870163-102870163 on Build GRCh38
rs372675994 Chr.10: 102870108-102870108 on Build GRCh38
rs747410528 Chr.10: 102870202-102870202 on Build GRCh38
rs201702937 Chr.10: 102870193-102870193 on Build GRCh38
rs527650795 Chr.10: 102870192-102870192 on Build GRCh38
rs762692824 Chr.10: 102870071-102870071 on Build GRCh38
rs868851524 Chr.10: 102870132-102870132 on Build GRCh38
rs755170490 Chr.10: 102870174-102870174 on Build GRCh38
rs373760875 Chr.10: 102870240-102870240 on Build GRCh38
rs111560337 Chr.10: 102870155-102870155 on Build GRCh38
rs964288395 Chr.10: 102870175-102870175 on Build GRCh38
rs781682494 Chr.10: 102870229-102870229 on Build GRCh38
rs752948206 Chr.10: 102870189-102870189 on Build GRCh38
rs17880345 Chr.10: 102870074-102870074 on Build GRCh38
rs766934575 Chr.10: 102870115-102870115 on Build GRCh38

View Full Product Details