Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134361425-134361650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
UniGene
Hs.426312
Gene Chromosome Location
Chr.3: 134355345-134375417 on Build GRCh38
Transcripts
NM_001278683.1 NM_001278685.1 NM_016201.3 XM_006713654.2 XM_011512881.1 XM_017006580.1 XM_017006581.1 XM_017006582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398283 c.1532G>T p.R511L Chr.3: 134361555-134361555 on Build GRCh38
4978517 c.1538G>A p.R513H Chr.3: 134361549-134361549 on Build GRCh38
1038644 c.1526G>A p.R509H Chr.3: 134361561-134361561 on Build GRCh38
3927355 c.1539C>T p.R513R Chr.3: 134361548-134361548 on Build GRCh38
4846288 c.1503G>A p.E501E Chr.3: 134361584-134361584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371070499 Chr.3: 134361524-134361524 on Build GRCh38
rs538657184 Chr.3: 134361624-134361624 on Build GRCh38
rs764237309 Chr.3: 134361612-134361612 on Build GRCh38
rs773003068 Chr.3: 134361579-134361579 on Build GRCh38
rs367811790 Chr.3: 134361568-134361568 on Build GRCh38
rs756267953 Chr.3: 134361519-134361519 on Build GRCh38
rs531119733 Chr.3: 134361450-134361450 on Build GRCh38
rs370345516 Chr.3: 134361625-134361625 on Build GRCh38
rs779215337 Chr.3: 134361630-134361630 on Build GRCh38
rs766297316 Chr.3: 134361595-134361595 on Build GRCh38
rs373578968 Chr.3: 134361554-134361554 on Build GRCh38
rs370001874 Chr.3: 134361542-134361542 on Build GRCh38
rs971833419 Chr.3: 134361613-134361613 on Build GRCh38
rs762778254 Chr.3: 134361590-134361590 on Build GRCh38
rs769326127 Chr.3: 134361576-134361576 on Build GRCh38
rs755039139 Chr.3: 134361494-134361494 on Build GRCh38
rs376131220 Chr.3: 134361490-134361490 on Build GRCh38
rs569708164 Chr.3: 134361611-134361611 on Build GRCh38
rs35993053 Chr.3: 134361528-134361528 on Build GRCh38
rs758995949 Chr.3: 134361501-134361501 on Build GRCh38
rs759361322 Chr.3: 134361607-134361607 on Build GRCh38
rs746390676 Chr.3: 134361556-134361556 on Build GRCh38
rs867847434 Chr.3: 134361515-134361515 on Build GRCh38
rs752863765 Chr.3: 134361504-134361504 on Build GRCh38
rs767540243 Chr.3: 134361608-134361608 on Build GRCh38
rs766350478 Chr.3: 134361489-134361489 on Build GRCh38
rs775979387 Chr.3: 134361570-134361570 on Build GRCh38
rs747748251 Chr.3: 134361571-134361571 on Build GRCh38
rs767526667 Chr.3: 134361499-134361499 on Build GRCh38
rs774393374 Chr.3: 134361601-134361601 on Build GRCh38
rs911614404 Chr.3: 134361540-134361540 on Build GRCh38

View Full Product Details