Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36514397-36514658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOAH
Entrez Gene ID
313
Gene Name
acyloxyacyl hydrolase
Gene Aliases
-
UniGene
Hs.488007
Gene Chromosome Location
Chr.7: 36512042-36724549 on Build GRCh38
Transcripts
NM_001177506.1 NM_001177507.1 NM_001637.3 XM_011515333.2 XM_011515334.2 XM_011515335.2 XM_011515336.2 XM_011515338.2 XM_011515342.2 XM_017012102.1 XM_017012103.1 XM_017012104.1 XM_017012105.1

SNP Detail

SNP ID Chromosome Location
rs916389616 Chr.7: 36514457-36514457 on Build GRCh38
rs140313251 Chr.7: 36514539-36514539 on Build GRCh38
rs759915099 Chr.7: 36514573-36514573 on Build GRCh38
rs939063042 Chr.7: 36514429-36514429 on Build GRCh38
rs145455591 Chr.7: 36514524-36514524 on Build GRCh38
rs934401200 Chr.7: 36514591-36514591 on Build GRCh38
rs985129754 Chr.7: 36514464-36514464 on Build GRCh38
rs55675361 Chr.7: 36514428-36514428 on Build GRCh38
rs749124951 Chr.7: 36514486-36514486 on Build GRCh38
rs910440635 Chr.7: 36514499-36514499 on Build GRCh38
rs547590795 Chr.7: 36514492-36514492 on Build GRCh38
rs774933077 Chr.7: 36514564-36514564 on Build GRCh38
rs1056253296 Chr.7: 36514456-36514456 on Build GRCh38
rs994060905 Chr.7: 36514520-36514520 on Build GRCh38
rs1019064295 Chr.7: 36514599-36514599 on Build GRCh38
rs923618463 Chr.7: 36514424-36514424 on Build GRCh38
rs527447203 Chr.7: 36514449-36514449 on Build GRCh38
rs898381064 Chr.7: 36514512-36514512 on Build GRCh38
rs753020203 Chr.7: 36514488-36514489 on Build GRCh38
rs768368152 Chr.7: 36514489-36514489 on Build GRCh38
rs766980761 Chr.7: 36514527-36514527 on Build GRCh38
rs549491477 Chr.7: 36514576-36514576 on Build GRCh38
rs1008924629 Chr.7: 36514578-36514578 on Build GRCh38
rs775375128 Chr.7: 36514477-36514477 on Build GRCh38
rs943037711 Chr.7: 36514510-36514510 on Build GRCh38
rs1038615110 Chr.7: 36514476-36514476 on Build GRCh38
rs373669032 Chr.7: 36514442-36514442 on Build GRCh38

View Full Product Details