Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134450707-134450980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B1
Entrez Gene ID
231
Gene Name
aldo-keto reductase family 1 member B
Gene Aliases
ADR, ALDR1, ALR2, AR
UniGene
Hs.521212
Gene Chromosome Location
Chr.7: 134442352-134459136 on Build GRCh38
Transcripts
NM_001628.2 XM_005250234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4780295 c.308A>G p.D103G Chr.7: 134450829-134450829 on Build GRCh38
4779486 c.284A>C p.K95T Chr.7: 134450853-134450853 on Build GRCh38
1312694 c.253G>A p.E85K Chr.7: 134450884-134450884 on Build GRCh38
81992 c.258G>A p.K86K Chr.7: 134450879-134450879 on Build GRCh38
5871873 c.331C>G p.H111D Chr.7: 134450806-134450806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034499939 Chr.7: 134450921-134450921 on Build GRCh38
rs775340161 Chr.7: 134450809-134450810 on Build GRCh38
rs765362829 Chr.7: 134450842-134450842 on Build GRCh38
rs201909884 Chr.7: 134450932-134450932 on Build GRCh38
rs775676690 Chr.7: 134450934-134450934 on Build GRCh38
rs879086858 Chr.7: 134450903-134450903 on Build GRCh38
rs1036613744 Chr.7: 134450890-134450890 on Build GRCh38
rs757019436 Chr.7: 134450761-134450761 on Build GRCh38
rs757291347 Chr.7: 134450769-134450769 on Build GRCh38
rs747726692 Chr.7: 134450906-134450906 on Build GRCh38
rs183614720 Chr.7: 134450770-134450770 on Build GRCh38
rs754746758 Chr.7: 134450941-134450943 on Build GRCh38
rs778577954 Chr.7: 134450900-134450900 on Build GRCh38
rs546024547 Chr.7: 134450756-134450756 on Build GRCh38
rs921994118 Chr.7: 134450849-134450849 on Build GRCh38
rs115638419 Chr.7: 134450829-134450829 on Build GRCh38
rs1013924469 Chr.7: 134450937-134450937 on Build GRCh38
rs771807907 Chr.7: 134450909-134450909 on Build GRCh38
rs750704319 Chr.7: 134450923-134450924 on Build GRCh38
rs773668747 Chr.7: 134450834-134450834 on Build GRCh38
rs752888001 Chr.7: 134450846-134450846 on Build GRCh38
rs200955767 Chr.7: 134450814-134450814 on Build GRCh38
rs755180808 Chr.7: 134450747-134450747 on Build GRCh38
rs977267597 Chr.7: 134450754-134450754 on Build GRCh38
rs559336093 Chr.7: 134450757-134450757 on Build GRCh38
rs116793843 Chr.7: 134450843-134450843 on Build GRCh38
rs777139745 Chr.7: 134450919-134450919 on Build GRCh38
rs2229542 Chr.7: 134450869-134450869 on Build GRCh38
rs781132505 Chr.7: 134450767-134450767 on Build GRCh38
rs764977993 Chr.7: 134450770-134450771 on Build GRCh38
rs143869945 Chr.7: 134450858-134450858 on Build GRCh38
rs927365130 Chr.7: 134450928-134450928 on Build GRCh38
rs547750546 Chr.7: 134450778-134450778 on Build GRCh38
rs890428946 Chr.7: 134450866-134450866 on Build GRCh38
rs754547262 Chr.7: 134450892-134450892 on Build GRCh38
rs764430673 Chr.7: 134450957-134450957 on Build GRCh38
rs759566331 Chr.7: 134450924-134450924 on Build GRCh38
rs751304426 Chr.7: 134450758-134450758 on Build GRCh38
rs13306161 Chr.7: 134450913-134450913 on Build GRCh38
rs148084524 Chr.7: 134450848-134450848 on Build GRCh38
rs761065461 Chr.7: 134450836-134450836 on Build GRCh38
rs932031834 Chr.7: 134450852-134450852 on Build GRCh38
rs766416869 Chr.7: 134450739-134450739 on Build GRCh38
rs2854850 Chr.7: 134450741-134450741 on Build GRCh38
rs777986468 Chr.7: 134450912-134450912 on Build GRCh38
rs771486099 Chr.7: 134450837-134450837 on Build GRCh38
rs5059 Chr.7: 134450904-134450904 on Build GRCh38
rs755435675 Chr.7: 134450832-134450832 on Build GRCh38
rs199863988 Chr.7: 134450798-134450798 on Build GRCh38
rs143736441 Chr.7: 134450845-134450845 on Build GRCh38
rs374449372 Chr.7: 134450952-134450952 on Build GRCh38
rs754080850 Chr.7: 134450812-134450812 on Build GRCh38
rs768419303 Chr.7: 134450799-134450799 on Build GRCh38
rs139738197 Chr.7: 134450891-134450891 on Build GRCh38
rs902261443 Chr.7: 134450746-134450746 on Build GRCh38
rs2852166 Chr.7: 134450742-134450742 on Build GRCh38
rs374068972 Chr.7: 134450839-134450839 on Build GRCh38
rs767183202 Chr.7: 134450857-134450857 on Build GRCh38
rs766919117 Chr.7: 134450784-134450784 on Build GRCh38
rs925845574 Chr.7: 134450780-134450780 on Build GRCh38
rs750357357 Chr.7: 134450771-134450771 on Build GRCh38
rs780206909 Chr.7: 134450886-134450886 on Build GRCh38
rs201357441 Chr.7: 134450816-134450816 on Build GRCh38

View Full Product Details