Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195285587-195285854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP2
Entrez Gene ID
23527
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
CENTB2, CNT-B2
UniGene
Hs.593373
Gene Chromosome Location
Chr.3: 195274736-195443176 on Build GRCh38
Transcripts
NM_012287.5 XM_006713557.3 XM_011512602.2 XM_011512603.2 XM_011512604.2 XM_011512605.2 XM_011512606.2 XM_017006046.1 XM_017006047.1 XM_017006048.1 XM_017006049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042761 c.2232G>T p.Q744H Chr.3: 195285800-195285800 on Build GRCh38
6365376 c.2222T>A p.L741H Chr.3: 195285810-195285810 on Build GRCh38
7400022 c.2236+1G>A p.? Chr.3: 195285795-195285795 on Build GRCh38
3992950 c.2235A>G p.P745P Chr.3: 195285797-195285797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974360415 Chr.3: 195285776-195285776 on Build GRCh38
rs950496538 Chr.3: 195285684-195285684 on Build GRCh38
rs768265659 Chr.3: 195285717-195285717 on Build GRCh38
rs563676667 Chr.3: 195285673-195285673 on Build GRCh38
rs192900704 Chr.3: 195285674-195285674 on Build GRCh38
rs911574603 Chr.3: 195285732-195285732 on Build GRCh38
rs987600492 Chr.3: 195285708-195285708 on Build GRCh38
rs928256739 Chr.3: 195285682-195285683 on Build GRCh38
rs777928557 Chr.3: 195285774-195285774 on Build GRCh38
rs936917970 Chr.3: 195285782-195285782 on Build GRCh38
rs973266164 Chr.3: 195285661-195285661 on Build GRCh38
rs867497776 Chr.3: 195285826-195285826 on Build GRCh38
rs961577329 Chr.3: 195285644-195285644 on Build GRCh38
rs76861098 Chr.3: 195285643-195285643 on Build GRCh38
rs545053669 Chr.3: 195285665-195285665 on Build GRCh38
rs779750471 Chr.3: 195285745-195285745 on Build GRCh38
rs770940146 Chr.3: 195285809-195285809 on Build GRCh38
rs1033626129 Chr.3: 195285610-195285610 on Build GRCh38
rs375440796 Chr.3: 195285777-195285777 on Build GRCh38
rs756347794 Chr.3: 195285772-195285772 on Build GRCh38
rs185816021 Chr.3: 195285749-195285749 on Build GRCh38

View Full Product Details