Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156940030-156940283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF11
Entrez Gene ID
9826
Gene Name
Rho guanine nucleotide exchange factor 11
Gene Aliases
GTRAP48, PDZ-RHOGEF
UniGene
Hs.516954
Gene Chromosome Location
Chr.1: 156934840-157046903 on Build GRCh38
Transcripts
NM_014784.3 NM_198236.2 XM_005245629.4 XM_005245633.4 XM_006711659.2 XM_006711660.3 XM_006711661.2 XM_006711662.2 XM_006711663.3 XM_006711665.3 XM_011510185.2 XM_011510186.2 XM_011510187.2 XM_011510189.2 XM_017002920.1 XM_017002921.1 XM_017002922.1 XM_017002923.1 XM_017002924.1 XM_017000459.1

Gene Symbol
LRRC71
Entrez Gene ID
149499
Gene Name
leucine rich repeat containing 71
Gene Aliases
C1orf92
UniGene
Hs.491734
Gene Chromosome Location
Chr.1: 156920608-156941208 on Build GRCh38
Transcripts
NM_014784.3 NM_198236.2 XM_005245629.4 XM_005245633.4 XM_006711659.2 XM_006711660.3 XM_006711661.2 XM_006711662.2 XM_006711663.3 XM_006711665.3 XM_011510185.2 XM_011510186.2 XM_011510187.2 XM_011510189.2 XM_017002920.1 XM_017002921.1 XM_017002922.1 XM_017002923.1 XM_017002924.1 XM_017000459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814973 c.1838T>A p.L613H Chr.1: 156940230-156940230 on Build GRCh38
3475554 c.1858G>A p.D620N Chr.1: 156940210-156940210 on Build GRCh38
6603928 c.3722C>T p.A1241V Chr.1: 156940218-156940218 on Build GRCh38
897474 c.3679G>A p.A1227T Chr.1: 156940261-156940261 on Build GRCh38
6260532 c.1861+8C>A p.? Chr.1: 156940199-156940199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377321852 Chr.1: 156940200-156940200 on Build GRCh38
rs969963836 Chr.1: 156940117-156940117 on Build GRCh38
rs200246881 Chr.1: 156940184-156940184 on Build GRCh38
rs144855685 Chr.1: 156940118-156940118 on Build GRCh38
rs968108752 Chr.1: 156940072-156940072 on Build GRCh38
rs540910645 Chr.1: 156940104-156940104 on Build GRCh38
rs780761122 Chr.1: 156940160-156940160 on Build GRCh38
rs1012082563 Chr.1: 156940255-156940255 on Build GRCh38
rs749172413 Chr.1: 156940169-156940169 on Build GRCh38
rs532547489 Chr.1: 156940154-156940154 on Build GRCh38
rs924145890 Chr.1: 156940084-156940084 on Build GRCh38
rs767953692 Chr.1: 156940249-156940249 on Build GRCh38
rs758817490 Chr.1: 156940183-156940183 on Build GRCh38
rs755537321 Chr.1: 156940164-156940164 on Build GRCh38
rs752258483 Chr.1: 156940251-156940251 on Build GRCh38
rs755283148 Chr.1: 156940240-156940240 on Build GRCh38
rs762137591 Chr.1: 156940204-156940204 on Build GRCh38
rs144803270 Chr.1: 156940228-156940228 on Build GRCh38
rs759414789 Chr.1: 156940139-156940139 on Build GRCh38
rs1022863610 Chr.1: 156940103-156940103 on Build GRCh38
rs377011765 Chr.1: 156940158-156940158 on Build GRCh38
rs763446124 Chr.1: 156940226-156940226 on Build GRCh38
rs909585483 Chr.1: 156940122-156940122 on Build GRCh38
rs201198340 Chr.1: 156940163-156940163 on Build GRCh38
rs548296599 Chr.1: 156940219-156940219 on Build GRCh38
rs745601196 Chr.1: 156940185-156940185 on Build GRCh38
rs376078328 Chr.1: 156940220-156940220 on Build GRCh38
rs942921043 Chr.1: 156940119-156940119 on Build GRCh38
rs1031901920 Chr.1: 156940071-156940071 on Build GRCh38
rs79719130 Chr.1: 156940157-156940157 on Build GRCh38
rs569602412 Chr.1: 156940229-156940229 on Build GRCh38
rs772016548 Chr.1: 156940196-156940196 on Build GRCh38
rs535684254 Chr.1: 156940178-156940178 on Build GRCh38
rs771062259 Chr.1: 156940176-156940176 on Build GRCh38
rs777394118 Chr.1: 156940166-156940166 on Build GRCh38
rs910764584 Chr.1: 156940227-156940227 on Build GRCh38
rs1014108390 Chr.1: 156940080-156940080 on Build GRCh38
rs959928269 Chr.1: 156940079-156940079 on Build GRCh38

View Full Product Details