Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122980367-122980616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP4
Entrez Gene ID
51329
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases
SFRS20, SR-25, SRp25, SRrp37
UniGene
Hs.103561
Gene Chromosome Location
Chr.12: 122980060-122982913 on Build GRCh38
Transcripts
NM_001002251.2 NM_001002252.2 NM_001278378.1 NM_001278379.1 NM_001278380.1 NM_016638.3 NM_018694.3 NR_103512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692470 c.127T>G p.S43A Chr.12: 122980514-122980514 on Build GRCh38
6268517 c.152G>T p.C51F Chr.12: 122980539-122980539 on Build GRCh38
6268516 c.152G>T p.C51F Chr.12: 122980539-122980539 on Build GRCh38
4663340 c.64G>A p.A22T Chr.12: 122980451-122980451 on Build GRCh38
6382502 c.151T>A p.C51S Chr.12: 122980538-122980538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769303922 Chr.12: 122980386-122980386 on Build GRCh38
rs201259682 Chr.12: 122980461-122980461 on Build GRCh38
rs750949380 Chr.12: 122980464-122980464 on Build GRCh38
rs750485699 Chr.12: 122980534-122980534 on Build GRCh38
rs758831948 Chr.12: 122980582-122980582 on Build GRCh38
rs748653483 Chr.12: 122980465-122980465 on Build GRCh38
rs767425808 Chr.12: 122980533-122980533 on Build GRCh38
rs768475946 Chr.12: 122980512-122980512 on Build GRCh38
rs758547682 Chr.12: 122980521-122980522 on Build GRCh38
rs555209961 Chr.12: 122980572-122980572 on Build GRCh38
rs775267501 Chr.12: 122980415-122980415 on Build GRCh38
rs983504506 Chr.12: 122980395-122980395 on Build GRCh38
rs747457503 Chr.12: 122980592-122980592 on Build GRCh38
rs750478780 Chr.12: 122980430-122980430 on Build GRCh38
rs754154557 Chr.12: 122980564-122980564 on Build GRCh38
rs762676950 Chr.12: 122980419-122980419 on Build GRCh38
rs878865242 Chr.12: 122980525-122980525 on Build GRCh38
rs372943330 Chr.12: 122980399-122980399 on Build GRCh38
rs568995765 Chr.12: 122980528-122980528 on Build GRCh38
rs536795391 Chr.12: 122980557-122980557 on Build GRCh38
rs778285529 Chr.12: 122980590-122980590 on Build GRCh38
rs775180773 Chr.12: 122980492-122980492 on Build GRCh38
rs911674877 Chr.12: 122980453-122980453 on Build GRCh38
rs375741783 Chr.12: 122980517-122980517 on Build GRCh38
rs765785252 Chr.12: 122980445-122980445 on Build GRCh38
rs1012461224 Chr.12: 122980554-122980554 on Build GRCh38
rs780369261 Chr.12: 122980434-122980434 on Build GRCh38
rs748685014 Chr.12: 122980396-122980396 on Build GRCh38
rs573457244 Chr.12: 122980578-122980578 on Build GRCh38
rs745308204 Chr.12: 122980479-122980479 on Build GRCh38
rs779503183 Chr.12: 122980596-122980596 on Build GRCh38
rs751561292 Chr.12: 122980424-122980424 on Build GRCh38
rs550658632 Chr.12: 122980468-122980468 on Build GRCh38
rs76875201 Chr.12: 122980475-122980475 on Build GRCh38
rs572174828 Chr.12: 122980390-122980390 on Build GRCh38
rs774170746 Chr.12: 122980515-122980515 on Build GRCh38
rs759132004 Chr.12: 122980408-122980408 on Build GRCh38
rs535334086 Chr.12: 122980555-122980555 on Build GRCh38
rs911815820 Chr.12: 122980490-122980490 on Build GRCh38
rs571246576 Chr.12: 122980420-122980420 on Build GRCh38
rs1054010058 Chr.12: 122980551-122980551 on Build GRCh38
rs755230941 Chr.12: 122980566-122980566 on Build GRCh38
rs375871449 Chr.12: 122980407-122980407 on Build GRCh38
rs779622157 Chr.12: 122980393-122980393 on Build GRCh38
rs748529387 Chr.12: 122980426-122980426 on Build GRCh38
rs762586641 Chr.12: 122980508-122980508 on Build GRCh38
rs767540748 Chr.12: 122980428-122980428 on Build GRCh38
rs1039125132 Chr.12: 122980526-122980526 on Build GRCh38
rs765721147 Chr.12: 122980569-122980569 on Build GRCh38
rs756298215 Chr.12: 122980432-122980432 on Build GRCh38
rs771938020 Chr.12: 122980541-122980541 on Build GRCh38
rs755461871 Chr.12: 122980443-122980443 on Build GRCh38
rs755684992 Chr.12: 122980594-122980594 on Build GRCh38
rs769449583 Chr.12: 122980485-122980485 on Build GRCh38
rs760899805 Chr.12: 122980540-122980540 on Build GRCh38
rs754103527 Chr.12: 122980438-122980438 on Build GRCh38
rs945771992 Chr.12: 122980480-122980480 on Build GRCh38
rs944347432 Chr.12: 122980498-122980498 on Build GRCh38
rs202084070 Chr.12: 122980409-122980409 on Build GRCh38
rs895320695 Chr.12: 122980552-122980552 on Build GRCh38

View Full Product Details