Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44177518-44177791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
UniGene
Hs.534517
Gene Chromosome Location
Chr.17: 44170706-44179083 on Build GRCh38
Transcripts
NM_080863.4 NR_049729.1 NR_049730.1

Gene Symbol
ASB16-AS1
Entrez Gene ID
339201
Gene Name
ASB16 antisense RNA 1
Gene Aliases
C17orf65
UniGene
Hs.656564
Gene Chromosome Location
Chr.17: 44175973-44186717 on Build GRCh38
Transcripts
NM_080863.4 NR_049729.1 NR_049730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1325042 c.1148T>C p.V383A Chr.17: 44177694-44177694 on Build GRCh38
979895 c.1077C>T p.C359C Chr.17: 44177623-44177623 on Build GRCh38
4402862 c.1089T>G p.P363P Chr.17: 44177635-44177635 on Build GRCh38
1610319 c.1078G>A p.A360T Chr.17: 44177624-44177624 on Build GRCh38
5913840 c.1099G>A p.E367K Chr.17: 44177645-44177645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036764808 Chr.17: 44177586-44177586 on Build GRCh38
rs755760868 Chr.17: 44177600-44177600 on Build GRCh38
rs760641432 Chr.17: 44177758-44177758 on Build GRCh38
rs555660936 Chr.17: 44177715-44177715 on Build GRCh38
rs746629092 Chr.17: 44177751-44177754 on Build GRCh38
rs551155706 Chr.17: 44177623-44177623 on Build GRCh38
rs747833042 Chr.17: 44177748-44177748 on Build GRCh38
rs777931851 Chr.17: 44177644-44177644 on Build GRCh38
rs776059267 Chr.17: 44177759-44177759 on Build GRCh38
rs745829254 Chr.17: 44177681-44177682 on Build GRCh38
rs376241371 Chr.17: 44177645-44177645 on Build GRCh38
rs749502395 Chr.17: 44177741-44177741 on Build GRCh38
rs778766270 Chr.17: 44177746-44177746 on Build GRCh38
rs774064652 Chr.17: 44177671-44177671 on Build GRCh38
rs768818953 Chr.17: 44177744-44177744 on Build GRCh38
rs768466732 Chr.17: 44177667-44177667 on Build GRCh38
rs761796864 Chr.17: 44177676-44177676 on Build GRCh38
rs775855615 Chr.17: 44177558-44177558 on Build GRCh38
rs752404314 Chr.17: 44177769-44177769 on Build GRCh38
rs759217833 Chr.17: 44177760-44177760 on Build GRCh38
rs771870273 Chr.17: 44177749-44177749 on Build GRCh38
rs141164758 Chr.17: 44177701-44177701 on Build GRCh38
rs369982324 Chr.17: 44177704-44177704 on Build GRCh38
rs749995678 Chr.17: 44177585-44177585 on Build GRCh38
rs375931013 Chr.17: 44177768-44177768 on Build GRCh38
rs781739786 Chr.17: 44177725-44177725 on Build GRCh38
rs772066282 Chr.17: 44177692-44177692 on Build GRCh38
rs906220218 Chr.17: 44177602-44177602 on Build GRCh38
rs1031740215 Chr.17: 44177742-44177742 on Build GRCh38
rs577810555 Chr.17: 44177747-44177747 on Build GRCh38
rs778033932 Chr.17: 44177624-44177624 on Build GRCh38
rs751931430 Chr.17: 44177716-44177716 on Build GRCh38
rs774304877 Chr.17: 44177570-44177570 on Build GRCh38
rs773086579 Chr.17: 44177754-44177754 on Build GRCh38
rs749502893 Chr.17: 44177657-44177657 on Build GRCh38
rs745873127 Chr.17: 44177640-44177640 on Build GRCh38
rs762750639 Chr.17: 44177562-44177562 on Build GRCh38
rs373010761 Chr.17: 44177578-44177578 on Build GRCh38
rs757335191 Chr.17: 44177660-44177660 on Build GRCh38
rs771881364 Chr.17: 44177718-44177722 on Build GRCh38
rs768304635 Chr.17: 44177753-44177759 on Build GRCh38
rs150322450 Chr.17: 44177719-44177719 on Build GRCh38
rs888586986 Chr.17: 44177557-44177557 on Build GRCh38
rs765899037 Chr.17: 44177702-44177702 on Build GRCh38
rs756436248 Chr.17: 44177614-44177614 on Build GRCh38
rs932867929 Chr.17: 44177543-44177543 on Build GRCh38
rs761299642 Chr.17: 44177758-44177759 on Build GRCh38
rs761697235 Chr.17: 44177574-44177574 on Build GRCh38
rs747525872 Chr.17: 44177626-44177626 on Build GRCh38
rs763815339 Chr.17: 44177566-44177566 on Build GRCh38
rs537801622 Chr.17: 44177727-44177727 on Build GRCh38
rs778903045 Chr.17: 44177662-44177663 on Build GRCh38
rs753976530 Chr.17: 44177643-44177644 on Build GRCh38
rs200302229 Chr.17: 44177621-44177621 on Build GRCh38
rs568967870 Chr.17: 44177544-44177544 on Build GRCh38
rs1005983704 Chr.17: 44177553-44177553 on Build GRCh38
rs147448281 Chr.17: 44177602-44177602 on Build GRCh38
rs775804321 Chr.17: 44177651-44177651 on Build GRCh38
rs779737997 Chr.17: 44177747-44177747 on Build GRCh38
rs928234223 Chr.17: 44177637-44177637 on Build GRCh38
rs776932156 Chr.17: 44177672-44177672 on Build GRCh38
rs3826412 Chr.17: 44177729-44177729 on Build GRCh38
rs192556911 Chr.17: 44177636-44177636 on Build GRCh38
rs959277945 Chr.17: 44177699-44177699 on Build GRCh38
rs776062431 Chr.17: 44177754-44177754 on Build GRCh38
rs376109929 Chr.17: 44177707-44177707 on Build GRCh38

View Full Product Details