Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32332262-32332501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7345070 c.903_904insA p.T302fs*3 Chr.13: 32332381-32332382 on Build GRCh38
469354 c.938C>A p.S313Y Chr.13: 32332416-32332416 on Build GRCh38
1747412 c.800G>A p.G267E Chr.13: 32332278-32332278 on Build GRCh38
3671267 c.841G>C p.D281H Chr.13: 32332319-32332319 on Build GRCh38
3468372 c.947G>A p.R316K Chr.13: 32332425-32332425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886040828 Chr.13: 32332405-32332405 on Build GRCh38
rs397507399 Chr.13: 32332303-32332303 on Build GRCh38
rs397507437 Chr.13: 32332476-32332477 on Build GRCh38
rs1052441923 Chr.13: 32332454-32332454 on Build GRCh38
rs886040759 Chr.13: 32332309-32332310 on Build GRCh38
rs80359068 Chr.13: 32332296-32332296 on Build GRCh38
rs80359085 Chr.13: 32332316-32332316 on Build GRCh38
rs431825364 Chr.13: 32332314-32332314 on Build GRCh38
rs886040773 Chr.13: 32332320-32332321 on Build GRCh38
rs80359234 Chr.13: 32332439-32332439 on Build GRCh38
rs1039490629 Chr.13: 32332397-32332397 on Build GRCh38
rs398122619 Chr.13: 32332446-32332446 on Build GRCh38
rs398122598 Chr.13: 32332292-32332292 on Build GRCh38
rs879255298 Chr.13: 32332367-32332367 on Build GRCh38
rs886040848 Chr.13: 32332448-32332451 on Build GRCh38
rs397508039 Chr.13: 32332396-32332397 on Build GRCh38
rs753352530 Chr.13: 32332359-32332359 on Build GRCh38
rs878853616 Chr.13: 32332376-32332376 on Build GRCh38
rs786201605 Chr.13: 32332297-32332297 on Build GRCh38
rs886040847 Chr.13: 32332446-32332449 on Build GRCh38
rs587781421 Chr.13: 32332315-32332315 on Build GRCh38
rs864622214 Chr.13: 32332447-32332447 on Build GRCh38
rs11571640 Chr.13: 32332442-32332442 on Build GRCh38
rs184408918 Chr.13: 32332396-32332396 on Build GRCh38
rs276174931 Chr.13: 32332473-32332474 on Build GRCh38
rs778907270 Chr.13: 32332406-32332406 on Build GRCh38
rs397508031 Chr.13: 32332379-32332380 on Build GRCh38
rs55939572 Chr.13: 32332434-32332434 on Build GRCh38
rs766173 Chr.13: 32332343-32332343 on Build GRCh38
rs890456700 Chr.13: 32332438-32332438 on Build GRCh38
rs768605944 Chr.13: 32332322-32332322 on Build GRCh38
rs876658861 Chr.13: 32332312-32332312 on Build GRCh38
rs397507428 Chr.13: 32332416-32332416 on Build GRCh38
rs765832839 Chr.13: 32332345-32332345 on Build GRCh38
rs886040776 Chr.13: 32332324-32332325 on Build GRCh38
rs886040846 Chr.13: 32332434-32332434 on Build GRCh38
rs776059220 Chr.13: 32332435-32332435 on Build GRCh38
rs876659359 Chr.13: 32332366-32332366 on Build GRCh38
rs28897706 Chr.13: 32332456-32332456 on Build GRCh38
rs28897705 Chr.13: 32332309-32332309 on Build GRCh38
rs397507435 Chr.13: 32332449-32332449 on Build GRCh38
rs61754140 Chr.13: 32332409-32332409 on Build GRCh38
rs80359770 Chr.13: 32332434-32332435 on Build GRCh38
rs80359111 Chr.13: 32332334-32332334 on Build GRCh38
rs397508033 Chr.13: 32332384-32332384 on Build GRCh38
rs587782691 Chr.13: 32332455-32332455 on Build GRCh38
rs730881507 Chr.13: 32332326-32332326 on Build GRCh38
rs876659712 Chr.13: 32332311-32332311 on Build GRCh38
rs397508060 Chr.13: 32332430-32332430 on Build GRCh38
rs774063092 Chr.13: 32332323-32332323 on Build GRCh38
rs886040824 Chr.13: 32332397-32332398 on Build GRCh38
rs397508009 Chr.13: 32332358-32332358 on Build GRCh38
rs79483201 Chr.13: 32332421-32332421 on Build GRCh38
rs886040946 Chr.13: 32332369-32332376 on Build GRCh38
rs80359735 Chr.13: 32332375-32332376 on Build GRCh38
rs879255473 Chr.13: 32332444-32332444 on Build GRCh38
rs561002197 Chr.13: 32332465-32332465 on Build GRCh38
rs886040815 Chr.13: 32332383-32332384 on Build GRCh38
rs80359169 Chr.13: 32332391-32332391 on Build GRCh38
rs276174927 Chr.13: 32332441-32332441 on Build GRCh38
rs886040795 Chr.13: 32332362-32332362 on Build GRCh38
rs781169306 Chr.13: 32332394-32332394 on Build GRCh38
rs876659899 Chr.13: 32332298-32332298 on Build GRCh38
rs397507400 Chr.13: 32332304-32332308 on Build GRCh38
rs876660385 Chr.13: 32332338-32332339 on Build GRCh38
rs80359738 Chr.13: 32332378-32332379 on Build GRCh38
rs397508066 Chr.13: 32332472-32332473 on Build GRCh38
rs886038059 Chr.13: 32332404-32332404 on Build GRCh38
rs886040843 Chr.13: 32332427-32332427 on Build GRCh38
rs771011424 Chr.13: 32332306-32332306 on Build GRCh38
rs753067824 Chr.13: 32332371-32332371 on Build GRCh38
rs765756715 Chr.13: 32332368-32332368 on Build GRCh38
rs921111470 Chr.13: 32332400-32332400 on Build GRCh38
rs80359253 Chr.13: 32332470-32332470 on Build GRCh38
rs80359806 Chr.13: 32332405-32332405 on Build GRCh38
rs886038058 Chr.13: 32332403-32332404 on Build GRCh38
rs587781431 Chr.13: 32332347-32332347 on Build GRCh38
rs867323565 Chr.13: 32332386-32332386 on Build GRCh38
rs80359158 Chr.13: 32332383-32332383 on Build GRCh38
rs786203887 Chr.13: 32332375-32332375 on Build GRCh38
rs80359778 Chr.13: 32332472-32332472 on Build GRCh38
rs80359777 Chr.13: 32332471-32332472 on Build GRCh38
rs730881508 Chr.13: 32332380-32332380 on Build GRCh38
rs45457795 Chr.13: 32332365-32332365 on Build GRCh38
rs778050506 Chr.13: 32332465-32332466 on Build GRCh38
rs80359097 Chr.13: 32332325-32332325 on Build GRCh38
rs80359755 Chr.13: 32332408-32332409 on Build GRCh38
rs781464949 Chr.13: 32332476-32332476 on Build GRCh38
rs886038057 Chr.13: 32332311-32332311 on Build GRCh38
rs80359088 Chr.13: 32332319-32332319 on Build GRCh38
rs80359242 Chr.13: 32332457-32332457 on Build GRCh38
rs757430441 Chr.13: 32332387-32332387 on Build GRCh38
rs751890878 Chr.13: 32332459-32332459 on Build GRCh38
rs276174914 Chr.13: 32332369-32332377 on Build GRCh38

View Full Product Details