Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66282954-66283227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH2
Entrez Gene ID
254263
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnil
UniGene
Hs.437072
Gene Chromosome Location
Chr.11: 66278201-66284214 on Build GRCh38
Transcripts
NM_182553.2 NR_073078.1 NR_073079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163591 c.234C>G p.F78L Chr.11: 66283070-66283070 on Build GRCh38
2167307 c.274G>T p.G92C Chr.11: 66283110-66283110 on Build GRCh38
7229600 c.243G>C p.M81I Chr.11: 66283079-66283079 on Build GRCh38
344053 c.231C>T p.L77L Chr.11: 66283067-66283067 on Build GRCh38
7191872 c.311+2T>C p.? Chr.11: 66283149-66283149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779029577 Chr.11: 66283150-66283150 on Build GRCh38
rs771043624 Chr.11: 66283020-66283020 on Build GRCh38
rs144316476 Chr.11: 66283118-66283118 on Build GRCh38
rs780280373 Chr.11: 66282999-66282999 on Build GRCh38
rs749494415 Chr.11: 66283003-66283003 on Build GRCh38
rs768049022 Chr.11: 66283061-66283061 on Build GRCh38
rs769736632 Chr.11: 66283025-66283025 on Build GRCh38
rs774361486 Chr.11: 66283006-66283006 on Build GRCh38
rs572293797 Chr.11: 66283026-66283026 on Build GRCh38
rs769570456 Chr.11: 66283175-66283175 on Build GRCh38
rs772577374 Chr.11: 66283167-66283168 on Build GRCh38
rs989068582 Chr.11: 66283014-66283015 on Build GRCh38
rs541034346 Chr.11: 66282986-66282986 on Build GRCh38
rs993497696 Chr.11: 66283169-66283169 on Build GRCh38
rs1041991429 Chr.11: 66282974-66282974 on Build GRCh38
rs1057317545 Chr.11: 66283046-66283046 on Build GRCh38
rs768722465 Chr.11: 66283004-66283004 on Build GRCh38
rs367948808 Chr.11: 66283162-66283162 on Build GRCh38
rs377680648 Chr.11: 66282996-66282996 on Build GRCh38
rs772311162 Chr.11: 66283167-66283167 on Build GRCh38
rs773647149 Chr.11: 66283189-66283189 on Build GRCh38
rs371719138 Chr.11: 66283173-66283173 on Build GRCh38
rs1013033386 Chr.11: 66283113-66283113 on Build GRCh38
rs374970154 Chr.11: 66283176-66283176 on Build GRCh38
rs780295864 Chr.11: 66283094-66283094 on Build GRCh38
rs746930606 Chr.11: 66283008-66283008 on Build GRCh38
rs781230148 Chr.11: 66283168-66283168 on Build GRCh38
rs200300899 Chr.11: 66283203-66283203 on Build GRCh38
rs763651869 Chr.11: 66283037-66283037 on Build GRCh38
rs762520972 Chr.11: 66283028-66283028 on Build GRCh38
rs149611469 Chr.11: 66283062-66283062 on Build GRCh38
rs755214124 Chr.11: 66283121-66283121 on Build GRCh38
rs759368401 Chr.11: 66283024-66283024 on Build GRCh38
rs71457709 Chr.11: 66283065-66283065 on Build GRCh38
rs974712612 Chr.11: 66282976-66282976 on Build GRCh38
rs768240277 Chr.11: 66283185-66283185 on Build GRCh38
rs776537588 Chr.11: 66283023-66283023 on Build GRCh38
rs372667746 Chr.11: 66283206-66283206 on Build GRCh38
rs762283302 Chr.11: 66283049-66283049 on Build GRCh38
rs756419596 Chr.11: 66283085-66283085 on Build GRCh38
rs981194721 Chr.11: 66283157-66283157 on Build GRCh38
rs751084280 Chr.11: 66283043-66283043 on Build GRCh38
rs762648829 Chr.11: 66283181-66283181 on Build GRCh38

View Full Product Details