Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74700398-74700665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2I
Entrez Gene ID
2969
Gene Name
general transcription factor IIi
Gene Aliases
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6
UniGene
Hs.647041
Gene Chromosome Location
Chr.7: 74657665-74760692 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

Gene Symbol
LOC101926943
Entrez Gene ID
101926943
Gene Name
uncharacterized LOC101926943
Gene Aliases
-
UniGene
Hs.654772
Gene Chromosome Location
Chr.7: 74688937-74728918 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

SNP Detail

SNP ID Chromosome Location
rs2301907 Chr.7: 74700517-74700517 on Build GRCh38
rs940505286 Chr.7: 74700582-74700582 on Build GRCh38
rs781868187 Chr.7: 74700452-74700452 on Build GRCh38
rs782469389 Chr.7: 74700463-74700463 on Build GRCh38
rs782021824 Chr.7: 74700566-74700566 on Build GRCh38
rs185375934 Chr.7: 74700576-74700576 on Build GRCh38
rs782702015 Chr.7: 74700628-74700628 on Build GRCh38
rs193037615 Chr.7: 74700472-74700472 on Build GRCh38
rs782653711 Chr.7: 74700542-74700542 on Build GRCh38
rs782685891 Chr.7: 74700578-74700579 on Build GRCh38
rs782239672 Chr.7: 74700471-74700471 on Build GRCh38
rs782005201 Chr.7: 74700476-74700476 on Build GRCh38
rs781943378 Chr.7: 74700594-74700594 on Build GRCh38
rs1020556819 Chr.7: 74700454-74700454 on Build GRCh38
rs781899988 Chr.7: 74700600-74700600 on Build GRCh38
rs147069558 Chr.7: 74700630-74700630 on Build GRCh38
rs782575518 Chr.7: 74700468-74700468 on Build GRCh38
rs966175842 Chr.7: 74700497-74700497 on Build GRCh38
rs782431867 Chr.7: 74700632-74700632 on Build GRCh38
rs782418739 Chr.7: 74700565-74700565 on Build GRCh38
rs141499295 Chr.7: 74700554-74700554 on Build GRCh38
rs782488840 Chr.7: 74700457-74700457 on Build GRCh38
rs782763502 Chr.7: 74700599-74700599 on Build GRCh38
rs782043969 Chr.7: 74700574-74700574 on Build GRCh38
rs782245383 Chr.7: 74700480-74700480 on Build GRCh38
rs782141408 Chr.7: 74700615-74700615 on Build GRCh38
rs782233836 Chr.7: 74700496-74700496 on Build GRCh38
rs782269443 Chr.7: 74700460-74700460 on Build GRCh38
rs782180402 Chr.7: 74700590-74700590 on Build GRCh38
rs1048924300 Chr.7: 74700575-74700575 on Build GRCh38
rs587593935 Chr.7: 74700505-74700505 on Build GRCh38

View Full Product Details