Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3120904-3121155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6968120 c.892C>T p.P298S Chr.19: 3120991-3120991 on Build GRCh38
6342221 c.917G>A p.R306Q Chr.19: 3121016-3121016 on Build GRCh38
2750407 c.1012C>T p.R338C Chr.19: 3121111-3121111 on Build GRCh38
6342222 c.1051C>T p.L351F Chr.19: 3121150-3121150 on Build GRCh38
6672109 c.914C>T p.A305V Chr.19: 3121013-3121013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772471502 Chr.19: 3121039-3121039 on Build GRCh38
rs747189004 Chr.19: 3121083-3121083 on Build GRCh38
rs145170690 Chr.19: 3121011-3121011 on Build GRCh38
rs367847109 Chr.19: 3120944-3120944 on Build GRCh38
rs997609345 Chr.19: 3120980-3120980 on Build GRCh38
rs781222908 Chr.19: 3120939-3120947 on Build GRCh38
rs963369532 Chr.19: 3120933-3120933 on Build GRCh38
rs919502623 Chr.19: 3120998-3120998 on Build GRCh38
rs772434708 Chr.19: 3121071-3121071 on Build GRCh38
rs752162469 Chr.19: 3121038-3121038 on Build GRCh38
rs745913963 Chr.19: 3120952-3120954 on Build GRCh38
rs750757769 Chr.19: 3121014-3121014 on Build GRCh38
rs776555145 Chr.19: 3121096-3121096 on Build GRCh38
rs386352309 Chr.19: 3121042-3121042 on Build GRCh38
rs753862594 Chr.19: 3121058-3121058 on Build GRCh38
rs928679789 Chr.19: 3121107-3121107 on Build GRCh38
rs201368515 Chr.19: 3121069-3121069 on Build GRCh38
rs139538868 Chr.19: 3121003-3121003 on Build GRCh38
rs777133920 Chr.19: 3121054-3121054 on Build GRCh38
rs555203104 Chr.19: 3121101-3121101 on Build GRCh38
rs752451221 Chr.19: 3121126-3121126 on Build GRCh38
rs777708537 Chr.19: 3120951-3120951 on Build GRCh38
rs770816627 Chr.19: 3120954-3120954 on Build GRCh38
rs539753774 Chr.19: 3121002-3121002 on Build GRCh38
rs778881101 Chr.19: 3121068-3121068 on Build GRCh38
rs780352816 Chr.19: 3121074-3121074 on Build GRCh38
rs773591539 Chr.19: 3121111-3121111 on Build GRCh38
rs762218250 Chr.19: 3121010-3121010 on Build GRCh38
rs767110044 Chr.19: 3121028-3121028 on Build GRCh38
rs763698658 Chr.19: 3121134-3121134 on Build GRCh38
rs749176772 Chr.19: 3120952-3120952 on Build GRCh38
rs760222003 Chr.19: 3121128-3121128 on Build GRCh38
rs775732619 Chr.19: 3120979-3120979 on Build GRCh38
rs372683011 Chr.19: 3120929-3120929 on Build GRCh38
rs759218880 Chr.19: 3121024-3121024 on Build GRCh38
rs746131264 Chr.19: 3120957-3120957 on Build GRCh38
rs763245208 Chr.19: 3121116-3121116 on Build GRCh38
rs770247248 Chr.19: 3121106-3121106 on Build GRCh38
rs375856122 Chr.19: 3121086-3121086 on Build GRCh38
rs760747005 Chr.19: 3120986-3120986 on Build GRCh38
rs547791756 Chr.19: 3120932-3120932 on Build GRCh38
rs150929326 Chr.19: 3121059-3121059 on Build GRCh38
rs928378285 Chr.19: 3121019-3121019 on Build GRCh38
rs755586030 Chr.19: 3121053-3121053 on Build GRCh38
rs140749796 Chr.19: 3121122-3121122 on Build GRCh38
rs894185941 Chr.19: 3121095-3121095 on Build GRCh38
rs770043233 Chr.19: 3120956-3120956 on Build GRCh38
rs772376276 Chr.19: 3120974-3120974 on Build GRCh38
rs566050701 Chr.19: 3120933-3120933 on Build GRCh38
rs973672769 Chr.19: 3120934-3120934 on Build GRCh38

View Full Product Details