Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3930300-3930557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A3
Entrez Gene ID
489
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Gene Aliases
SERCA3
UniGene
Hs.513870
Gene Chromosome Location
Chr.17: 3923869-3964464 on Build GRCh38
Transcripts
NM_005173.3 NM_174953.2 NM_174954.2 NM_174955.2 NM_174956.2 NM_174957.2 NM_174958.2 XM_011523881.2 XM_011523882.2 XM_011523884.2 XM_011523885.1 XM_011523888.2 XM_011523889.1 XM_017024692.1 XM_017024693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400230 c.2611-1G>C p.? Chr.17: 3930435-3930435 on Build GRCh38
1709583 c.2705C>T p.S902F Chr.17: 3930340-3930340 on Build GRCh38
6002432 c.2710C>T p.L904F Chr.17: 3930335-3930335 on Build GRCh38
4874231 c.210C>T p.I70I Chr.17: 3930387-3930387 on Build GRCh38
1660689 c.2707G>A p.V903M Chr.17: 3930338-3930338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776296096 Chr.17: 3930445-3930446 on Build GRCh38
rs141235119 Chr.17: 3930371-3930371 on Build GRCh38
rs902458745 Chr.17: 3930496-3930496 on Build GRCh38
rs749378471 Chr.17: 3930440-3930440 on Build GRCh38
rs750399072 Chr.17: 3930460-3930460 on Build GRCh38
rs768216161 Chr.17: 3930417-3930417 on Build GRCh38
rs372639683 Chr.17: 3930470-3930470 on Build GRCh38
rs960549901 Chr.17: 3930491-3930491 on Build GRCh38
rs749920008 Chr.17: 3930410-3930410 on Build GRCh38
rs10153291 Chr.17: 3930446-3930446 on Build GRCh38
rs781581788 Chr.17: 3930429-3930429 on Build GRCh38
rs754592975 Chr.17: 3930471-3930471 on Build GRCh38
rs762361959 Chr.17: 3930370-3930370 on Build GRCh38
rs777570900 Chr.17: 3930321-3930321 on Build GRCh38
rs147136003 Chr.17: 3930394-3930394 on Build GRCh38
rs761653041 Chr.17: 3930450-3930450 on Build GRCh38
rs368619808 Chr.17: 3930413-3930413 on Build GRCh38
rs1030807036 Chr.17: 3930487-3930487 on Build GRCh38
rs368016888 Chr.17: 3930333-3930333 on Build GRCh38
rs149912039 Chr.17: 3930392-3930392 on Build GRCh38
rs943429021 Chr.17: 3930490-3930490 on Build GRCh38
rs150775059 Chr.17: 3930377-3930377 on Build GRCh38
rs999341143 Chr.17: 3930526-3930526 on Build GRCh38
rs765154035 Chr.17: 3930453-3930453 on Build GRCh38
rs138759313 Chr.17: 3930354-3930354 on Build GRCh38
rs769104610 Chr.17: 3930365-3930365 on Build GRCh38
rs778396748 Chr.17: 3930485-3930485 on Build GRCh38
rs766499633 Chr.17: 3930384-3930384 on Build GRCh38
rs1053303229 Chr.17: 3930519-3930519 on Build GRCh38
rs536704191 Chr.17: 3930443-3930443 on Build GRCh38
rs747329092 Chr.17: 3930364-3930364 on Build GRCh38
rs761483577 Chr.17: 3930447-3930448 on Build GRCh38
rs773917238 Chr.17: 3930438-3930438 on Build GRCh38
rs200098007 Chr.17: 3930465-3930465 on Build GRCh38
rs757824255 Chr.17: 3930462-3930462 on Build GRCh38
rs367863718 Chr.17: 3930461-3930461 on Build GRCh38
rs1011232227 Chr.17: 3930527-3930527 on Build GRCh38
rs889793046 Chr.17: 3930464-3930464 on Build GRCh38
rs748512735 Chr.17: 3930477-3930477 on Build GRCh38
rs376219456 Chr.17: 3930437-3930437 on Build GRCh38
rs779812985 Chr.17: 3930345-3930345 on Build GRCh38
rs757204092 Chr.17: 3930331-3930331 on Build GRCh38
rs763896125 Chr.17: 3930391-3930391 on Build GRCh38
rs73330907 Chr.17: 3930386-3930386 on Build GRCh38
rs753858633 Chr.17: 3930325-3930325 on Build GRCh38
rs751899273 Chr.17: 3930458-3930459 on Build GRCh38
rs776982530 Chr.17: 3930367-3930367 on Build GRCh38
rs536271392 Chr.17: 3930492-3930492 on Build GRCh38
rs112214270 Chr.17: 3930414-3930414 on Build GRCh38
rs771653439 Chr.17: 3930339-3930339 on Build GRCh38
rs553373438 Chr.17: 3930500-3930500 on Build GRCh38
rs756616357 Chr.17: 3930479-3930479 on Build GRCh38
rs778352144 Chr.17: 3930332-3930332 on Build GRCh38
rs771088134 Chr.17: 3930441-3930441 on Build GRCh38
rs991948215 Chr.17: 3930495-3930495 on Build GRCh38
rs778832579 Chr.17: 3930403-3930403 on Build GRCh38
rs760360769 Chr.17: 3930387-3930387 on Build GRCh38
rs375431156 Chr.17: 3930393-3930393 on Build GRCh38
rs759760922 Chr.17: 3930445-3930445 on Build GRCh38
rs780723294 Chr.17: 3930476-3930476 on Build GRCh38
rs1011158596 Chr.17: 3930335-3930335 on Build GRCh38

View Full Product Details