Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47658802-47659061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf65
Entrez Gene ID
401335
Gene Name
chromosome 7 open reading frame 65
Gene Aliases
-
UniGene
Hs.520816
Gene Chromosome Location
Chr.7: 47655244-47661648 on Build GRCh38
Transcripts
NM_001123065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319129 c.185G>C p.R62T Chr.7: 47658957-47658957 on Build GRCh38
3263623 c.211A>T p.S71C Chr.7: 47658983-47658983 on Build GRCh38
3412105 c.223C>A p.L75I Chr.7: 47658995-47658995 on Build GRCh38
6873375 c.147G>A p.E49E Chr.7: 47658919-47658919 on Build GRCh38
7196106 c.219G>A p.E73E Chr.7: 47658991-47658991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370939216 Chr.7: 47658998-47658998 on Build GRCh38
rs756889678 Chr.7: 47659026-47659026 on Build GRCh38
rs370571842 Chr.7: 47658860-47658860 on Build GRCh38
rs375333624 Chr.7: 47658984-47658984 on Build GRCh38
rs777313070 Chr.7: 47658837-47658837 on Build GRCh38
rs779809697 Chr.7: 47658946-47658947 on Build GRCh38
rs201413304 Chr.7: 47658909-47658909 on Build GRCh38
rs371871055 Chr.7: 47658963-47658963 on Build GRCh38
rs777715066 Chr.7: 47658871-47658871 on Build GRCh38
rs765570303 Chr.7: 47658995-47658995 on Build GRCh38
rs888200446 Chr.7: 47659035-47659035 on Build GRCh38
rs775594736 Chr.7: 47658888-47658888 on Build GRCh38
rs994997270 Chr.7: 47658991-47658991 on Build GRCh38
rs370209990 Chr.7: 47658952-47658952 on Build GRCh38
rs375422953 Chr.7: 47658911-47658911 on Build GRCh38
rs115611995 Chr.7: 47659009-47659009 on Build GRCh38
rs770674705 Chr.7: 47658970-47658970 on Build GRCh38
rs776904379 Chr.7: 47658993-47658993 on Build GRCh38
rs571868304 Chr.7: 47658933-47658933 on Build GRCh38
rs769532649 Chr.7: 47658869-47658869 on Build GRCh38
rs191232227 Chr.7: 47658843-47658843 on Build GRCh38
rs768705655 Chr.7: 47658992-47658992 on Build GRCh38
rs772097438 Chr.7: 47658879-47658879 on Build GRCh38
rs772002000 Chr.7: 47658979-47658979 on Build GRCh38
rs751162715 Chr.7: 47658840-47658840 on Build GRCh38
rs763669231 Chr.7: 47659023-47659023 on Build GRCh38
rs944971853 Chr.7: 47658960-47658960 on Build GRCh38
rs774223449 Chr.7: 47658874-47658874 on Build GRCh38
rs778751610 Chr.7: 47658971-47658971 on Build GRCh38
rs760688238 Chr.7: 47658987-47658987 on Build GRCh38
rs745591002 Chr.7: 47658869-47658869 on Build GRCh38
rs759489502 Chr.7: 47658876-47658876 on Build GRCh38
rs368781043 Chr.7: 47658936-47658936 on Build GRCh38
rs76203945 Chr.7: 47658962-47658962 on Build GRCh38
rs992732636 Chr.7: 47658835-47658835 on Build GRCh38
rs377259928 Chr.7: 47658890-47658890 on Build GRCh38
rs534484572 Chr.7: 47658953-47658953 on Build GRCh38
rs551789387 Chr.7: 47658872-47658872 on Build GRCh38
rs755754059 Chr.7: 47659021-47659021 on Build GRCh38
rs375012975 Chr.7: 47658957-47658957 on Build GRCh38
rs1001905395 Chr.7: 47658908-47658908 on Build GRCh38
rs766851688 Chr.7: 47659003-47659003 on Build GRCh38
rs765627902 Chr.7: 47658943-47658943 on Build GRCh38
rs745818820 Chr.7: 47658972-47658972 on Build GRCh38
rs750750036 Chr.7: 47658997-47658997 on Build GRCh38
rs753596399 Chr.7: 47659024-47659024 on Build GRCh38
rs372865348 Chr.7: 47658873-47658873 on Build GRCh38
rs556116108 Chr.7: 47658927-47658927 on Build GRCh38
rs868112972 Chr.7: 47659010-47659010 on Build GRCh38
rs778608178 Chr.7: 47659031-47659031 on Build GRCh38
rs764230955 Chr.7: 47658892-47658892 on Build GRCh38
rs763374504 Chr.7: 47659002-47659002 on Build GRCh38

View Full Product Details