Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2864624-2864863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXM1
Entrez Gene ID
2305
Gene Name
forkhead box M1
Gene Aliases
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2857681-2877155 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520932.1 XM_011520933.1 XM_011520934.2 XM_011520935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666400 c.1030C>T p.R344* Chr.12: 2864743-2864743 on Build GRCh38
7225459 c.1052G>T p.R351L Chr.12: 2864721-2864721 on Build GRCh38
6666401 c.1086C>T p.G362G Chr.12: 2864687-2864687 on Build GRCh38
375299 c.1071C>T p.T357T Chr.12: 2864702-2864702 on Build GRCh38
4685419 c.1077C>T p.L359L Chr.12: 2864696-2864696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773580345 Chr.12: 2864671-2864671 on Build GRCh38
rs376160082 Chr.12: 2864701-2864701 on Build GRCh38
rs750087295 Chr.12: 2864691-2864691 on Build GRCh38
rs763565934 Chr.12: 2864672-2864672 on Build GRCh38
rs752962627 Chr.12: 2864743-2864743 on Build GRCh38
rs931802936 Chr.12: 2864647-2864647 on Build GRCh38
rs767649969 Chr.12: 2864775-2864775 on Build GRCh38
rs900046597 Chr.12: 2864739-2864739 on Build GRCh38
rs759633706 Chr.12: 2864698-2864698 on Build GRCh38
rs757991234 Chr.12: 2864729-2864729 on Build GRCh38
rs749681691 Chr.12: 2864767-2864767 on Build GRCh38
rs143532828 Chr.12: 2864721-2864721 on Build GRCh38
rs559378752 Chr.12: 2864801-2864801 on Build GRCh38
rs951600700 Chr.12: 2864834-2864834 on Build GRCh38
rs974218815 Chr.12: 2864727-2864727 on Build GRCh38
rs200486055 Chr.12: 2864722-2864722 on Build GRCh38
rs35610962 Chr.12: 2864650-2864651 on Build GRCh38
rs577007302 Chr.12: 2864724-2864724 on Build GRCh38
rs761005665 Chr.12: 2864796-2864796 on Build GRCh38
rs376822769 Chr.12: 2864656-2864656 on Build GRCh38
rs80022864 Chr.12: 2864716-2864716 on Build GRCh38
rs755853763 Chr.12: 2864702-2864702 on Build GRCh38
rs140482241 Chr.12: 2864683-2864683 on Build GRCh38
rs552495230 Chr.12: 2864695-2864695 on Build GRCh38
rs748399754 Chr.12: 2864661-2864661 on Build GRCh38
rs1029952651 Chr.12: 2864778-2864778 on Build GRCh38
rs759541711 Chr.12: 2864771-2864771 on Build GRCh38
rs775560652 Chr.12: 2864786-2864786 on Build GRCh38
rs779569686 Chr.12: 2864738-2864738 on Build GRCh38
rs139551994 Chr.12: 2864827-2864827 on Build GRCh38
rs774849581 Chr.12: 2864770-2864770 on Build GRCh38
rs34100819 Chr.12: 2864696-2864696 on Build GRCh38
rs746734860 Chr.12: 2864742-2864742 on Build GRCh38
rs962805049 Chr.12: 2864814-2864814 on Build GRCh38
rs1023683014 Chr.12: 2864723-2864723 on Build GRCh38
rs369795053 Chr.12: 2864665-2864665 on Build GRCh38
rs1030950664 Chr.12: 2864657-2864657 on Build GRCh38
rs542082303 Chr.12: 2864833-2864833 on Build GRCh38
rs146299415 Chr.12: 2864725-2864725 on Build GRCh38
rs774372195 Chr.12: 2864686-2864686 on Build GRCh38
rs745405351 Chr.12: 2864648-2864648 on Build GRCh38
rs982977051 Chr.12: 2864797-2864797 on Build GRCh38
rs771375590 Chr.12: 2864769-2864769 on Build GRCh38
rs771804320 Chr.12: 2864652-2864652 on Build GRCh38
rs767668214 Chr.12: 2864699-2864699 on Build GRCh38
rs982968201 Chr.12: 2864837-2864837 on Build GRCh38

View Full Product Details