Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7766481-7766734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4M
Entrez Gene ID
10332
Gene Name
C-type lectin domain family 4 member M
Gene Aliases
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
UniGene
Hs.421437
Gene Chromosome Location
Chr.19: 7763052-7770559 on Build GRCh38
Transcripts
NM_001144904.1 NM_001144905.1 NM_001144906.1 NM_001144907.1 NM_001144908.1 NM_001144909.1 NM_001144910.1 NM_001144911.1 NM_014257.4 NR_026707.1 NR_026708.1 NR_026709.1 XM_006722611.2 XM_006722612.3 XM_006722613.3 XM_006722614.3 XM_006722615.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4430045 c.843C>T p.F281F Chr.19: 7766714-7766714 on Build GRCh38
3893921 c.785-1G>A p.? Chr.19: 7766655-7766655 on Build GRCh38
5391168 c.653C>T p.S218F Chr.19: 7766725-7766725 on Build GRCh38
3893922 c.829G>A p.G277R Chr.19: 7766700-7766700 on Build GRCh38
5722802 c.834C>T p.N278N Chr.19: 7766705-7766705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748499359 Chr.19: 7766711-7766711 on Build GRCh38
rs571493120 Chr.19: 7766585-7766585 on Build GRCh38
rs187212560 Chr.19: 7766576-7766576 on Build GRCh38
rs765624262 Chr.19: 7766564-7766564 on Build GRCh38
rs780003860 Chr.19: 7766605-7766605 on Build GRCh38
rs475896 Chr.19: 7766543-7766543 on Build GRCh38
rs887060893 Chr.19: 7766646-7766646 on Build GRCh38
rs747108025 Chr.19: 7766670-7766670 on Build GRCh38
rs924891757 Chr.19: 7766527-7766527 on Build GRCh38
rs961223770 Chr.19: 7766551-7766551 on Build GRCh38
rs144617264 Chr.19: 7766668-7766668 on Build GRCh38
rs759174550 Chr.19: 7766649-7766649 on Build GRCh38
rs752539952 Chr.19: 7766658-7766658 on Build GRCh38
rs768784019 Chr.19: 7766618-7766618 on Build GRCh38
rs763382335 Chr.19: 7766635-7766635 on Build GRCh38
rs755162497 Chr.19: 7766672-7766672 on Build GRCh38
rs372984727 Chr.19: 7766623-7766623 on Build GRCh38
rs773762465 Chr.19: 7766647-7766647 on Build GRCh38
rs926985004 Chr.19: 7766606-7766606 on Build GRCh38
rs912238401 Chr.19: 7766566-7766566 on Build GRCh38
rs200725157 Chr.19: 7766693-7766693 on Build GRCh38
rs921088150 Chr.19: 7766624-7766624 on Build GRCh38
rs770083895 Chr.19: 7766714-7766714 on Build GRCh38
rs762205399 Chr.19: 7766628-7766628 on Build GRCh38
rs747216154 Chr.19: 7766613-7766613 on Build GRCh38
rs763996747 Chr.19: 7766660-7766660 on Build GRCh38
rs140767813 Chr.19: 7766659-7766659 on Build GRCh38
rs537354631 Chr.19: 7766651-7766651 on Build GRCh38
rs770281758 Chr.19: 7766633-7766633 on Build GRCh38
rs972547717 Chr.19: 7766556-7766556 on Build GRCh38
rs980219607 Chr.19: 7766596-7766596 on Build GRCh38
rs267605768 Chr.19: 7766701-7766701 on Build GRCh38
rs765844597 Chr.19: 7766642-7766642 on Build GRCh38
rs1023502164 Chr.19: 7766671-7766671 on Build GRCh38
rs773765213 Chr.19: 7766634-7766634 on Build GRCh38
rs753745106 Chr.19: 7766661-7766661 on Build GRCh38
rs559487513 Chr.19: 7766536-7766536 on Build GRCh38
rs377334555 Chr.19: 7766667-7766667 on Build GRCh38
rs781459490 Chr.19: 7766687-7766687 on Build GRCh38

View Full Product Details