Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84422363-84422620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C2
Entrez Gene ID
9914
Gene Name
ATPase secretory pathway Ca2+ transporting 2
Gene Aliases
SPCA2
UniGene
Hs.6168
Gene Chromosome Location
Chr.16: 84368523-84465777 on Build GRCh38
Transcripts
NM_001286527.2 NM_001291454.1 NM_014861.3 XM_011523486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555958 c.764G>T p.G255V Chr.16: 84422529-84422529 on Build GRCh38
3512987 c.723C>T p.T241T Chr.16: 84422488-84422488 on Build GRCh38
6050296 c.625-5C>A p.? Chr.16: 84422385-84422385 on Build GRCh38
5851629 c.631G>A p.D211N Chr.16: 84422396-84422396 on Build GRCh38
6050297 c.625-3T>G p.? Chr.16: 84422387-84422387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553518485 Chr.16: 84422501-84422501 on Build GRCh38
rs74038217 Chr.16: 84422532-84422532 on Build GRCh38
rs746908431 Chr.16: 84422425-84422425 on Build GRCh38
rs773529940 Chr.16: 84422546-84422546 on Build GRCh38
rs199681456 Chr.16: 84422583-84422583 on Build GRCh38
rs751902189 Chr.16: 84422426-84422426 on Build GRCh38
rs748293046 Chr.16: 84422539-84422539 on Build GRCh38
rs200198885 Chr.16: 84422561-84422561 on Build GRCh38
rs766403810 Chr.16: 84422440-84422440 on Build GRCh38
rs768764728 Chr.16: 84422475-84422475 on Build GRCh38
rs755616428 Chr.16: 84422459-84422465 on Build GRCh38
rs527646596 Chr.16: 84422385-84422385 on Build GRCh38
rs374924150 Chr.16: 84422585-84422585 on Build GRCh38
rs747625551 Chr.16: 84422460-84422460 on Build GRCh38
rs773298581 Chr.16: 84422496-84422496 on Build GRCh38
rs1054401005 Chr.16: 84422577-84422577 on Build GRCh38
rs200348030 Chr.16: 84422408-84422408 on Build GRCh38
rs753850251 Chr.16: 84422512-84422512 on Build GRCh38
rs760574352 Chr.16: 84422500-84422500 on Build GRCh38
rs377548282 Chr.16: 84422573-84422573 on Build GRCh38
rs538038991 Chr.16: 84422438-84422438 on Build GRCh38
rs757917806 Chr.16: 84422521-84422521 on Build GRCh38
rs748832344 Chr.16: 84422399-84422399 on Build GRCh38
rs974321077 Chr.16: 84422466-84422466 on Build GRCh38
rs372267356 Chr.16: 84422471-84422471 on Build GRCh38
rs202138543 Chr.16: 84422394-84422394 on Build GRCh38
rs754440299 Chr.16: 84422401-84422401 on Build GRCh38
rs750244482 Chr.16: 84422435-84422435 on Build GRCh38
rs751778433 Chr.16: 84422381-84422381 on Build GRCh38
rs368931246 Chr.16: 84422523-84422523 on Build GRCh38
rs553814658 Chr.16: 84422450-84422453 on Build GRCh38
rs199657072 Chr.16: 84422542-84422542 on Build GRCh38
rs376844362 Chr.16: 84422488-84422488 on Build GRCh38
rs764145528 Chr.16: 84422427-84422427 on Build GRCh38
rs377084195 Chr.16: 84422452-84422452 on Build GRCh38
rs754690455 Chr.16: 84422448-84422451 on Build GRCh38
rs533777907 Chr.16: 84422497-84422497 on Build GRCh38
rs369777857 Chr.16: 84422470-84422470 on Build GRCh38
rs549313591 Chr.16: 84422416-84422416 on Build GRCh38
rs759581204 Chr.16: 84422513-84422513 on Build GRCh38
rs772123494 Chr.16: 84422540-84422540 on Build GRCh38
rs200739405 Chr.16: 84422537-84422537 on Build GRCh38
rs771141399 Chr.16: 84422556-84422556 on Build GRCh38
rs374592085 Chr.16: 84422564-84422564 on Build GRCh38
rs754579976 Chr.16: 84422455-84422455 on Build GRCh38
rs376245920 Chr.16: 84422557-84422557 on Build GRCh38
rs774435521 Chr.16: 84422478-84422478 on Build GRCh38
rs777512574 Chr.16: 84422463-84422463 on Build GRCh38
rs371870750 Chr.16: 84422424-84422424 on Build GRCh38
rs184593151 Chr.16: 84422426-84422426 on Build GRCh38
rs752270464 Chr.16: 84422519-84422519 on Build GRCh38
rs200504983 Chr.16: 84422395-84422395 on Build GRCh38
rs769165218 Chr.16: 84422536-84422536 on Build GRCh38
rs761822961 Chr.16: 84422484-84422484 on Build GRCh38
rs749604077 Chr.16: 84422535-84422535 on Build GRCh38
rs765272369 Chr.16: 84422572-84422572 on Build GRCh38
rs752178731 Chr.16: 84422454-84422455 on Build GRCh38
rs762053691 Chr.16: 84422433-84422433 on Build GRCh38
rs763492589 Chr.16: 84422582-84422582 on Build GRCh38
rs1036302778 Chr.16: 84422530-84422530 on Build GRCh38
rs757101778 Chr.16: 84422383-84422383 on Build GRCh38
rs908555611 Chr.16: 84422498-84422498 on Build GRCh38
rs765221214 Chr.16: 84422516-84422516 on Build GRCh38
rs778531515 Chr.16: 84422459-84422459 on Build GRCh38
rs774606455 Chr.16: 84422430-84422430 on Build GRCh38
rs373166793 Chr.16: 84422526-84422526 on Build GRCh38
rs778762505 Chr.16: 84422402-84422402 on Build GRCh38
rs760772928 Chr.16: 84422547-84422547 on Build GRCh38
rs780251082 Chr.16: 84422534-84422534 on Build GRCh38
rs762610988 Chr.16: 84422579-84422579 on Build GRCh38
rs372288677 Chr.16: 84422434-84422434 on Build GRCh38
rs758150748 Chr.16: 84422462-84422462 on Build GRCh38

View Full Product Details