Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123762403-123762646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
UniGene
Hs.622654
Gene Chromosome Location
Chr.12: 123761995-123935720 on Build GRCh38
Transcripts
NM_207437.3 XM_005253555.3 XM_011538014.2 XM_011538015.2 XM_011538016.2 XM_011538017.2 XM_011538020.2 XM_017018960.1 XM_017018961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
936495 c.13G>A p.E5K Chr.12: 123762532-123762532 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748754034 Chr.12: 123762565-123762565 on Build GRCh38
rs748353395 Chr.12: 123762492-123762492 on Build GRCh38
rs913836272 Chr.12: 123762618-123762618 on Build GRCh38
rs778077713 Chr.12: 123762581-123762581 on Build GRCh38
rs760053392 Chr.12: 123762599-123762599 on Build GRCh38
rs777153512 Chr.12: 123762523-123762523 on Build GRCh38
rs755899458 Chr.12: 123762549-123762549 on Build GRCh38
rs558793917 Chr.12: 123762624-123762624 on Build GRCh38
rs761450398 Chr.12: 123762536-123762536 on Build GRCh38
rs373084498 Chr.12: 123762541-123762541 on Build GRCh38
rs995607490 Chr.12: 123762486-123762486 on Build GRCh38
rs758179532 Chr.12: 123762596-123762596 on Build GRCh38
rs755052875 Chr.12: 123762562-123762562 on Build GRCh38
rs773700732 Chr.12: 123762515-123762515 on Build GRCh38
rs371020253 Chr.12: 123762504-123762504 on Build GRCh38
rs751252736 Chr.12: 123762534-123762534 on Build GRCh38
rs764881946 Chr.12: 123762518-123762518 on Build GRCh38
rs772438964 Chr.12: 123762580-123762580 on Build GRCh38
rs889738912 Chr.12: 123762544-123762544 on Build GRCh38
rs535557000 Chr.12: 123762424-123762424 on Build GRCh38
rs377606125 Chr.12: 123762568-123762568 on Build GRCh38
rs987988489 Chr.12: 123762610-123762610 on Build GRCh38
rs995803071 Chr.12: 123762451-123762451 on Build GRCh38
rs1047359013 Chr.12: 123762512-123762512 on Build GRCh38
rs750231257 Chr.12: 123762548-123762548 on Build GRCh38
rs553708367 Chr.12: 123762447-123762447 on Build GRCh38
rs967309880 Chr.12: 123762621-123762621 on Build GRCh38
rs748443328 Chr.12: 123762576-123762576 on Build GRCh38
rs763635181 Chr.12: 123762530-123762530 on Build GRCh38
rs1039905885 Chr.12: 123762432-123762432 on Build GRCh38
rs969201290 Chr.12: 123762531-123762531 on Build GRCh38
rs753792797 Chr.12: 123762560-123762560 on Build GRCh38
rs966555227 Chr.12: 123762570-123762570 on Build GRCh38
rs779880841 Chr.12: 123762555-123762555 on Build GRCh38
rs572124409 Chr.12: 123762527-123762527 on Build GRCh38
rs771343451 Chr.12: 123762520-123762520 on Build GRCh38
rs752390916 Chr.12: 123762591-123762591 on Build GRCh38

View Full Product Details