Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154699895-154700168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB3
Entrez Gene ID
139716
Gene Name
GRB2 associated binding protein 3
Gene Aliases
-
UniGene
Hs.496982
Gene Chromosome Location
Chr.X: 154675249-154751583 on Build GRCh38
Transcripts
NM_001081573.2 NM_001282283.1 NM_080612.3 NR_104114.1 XM_005274648.1 XM_006724804.1 XM_011531103.1 XM_011531105.1 XM_011531106.1 XM_017029276.1

SNP Detail

SNP ID Chromosome Location
rs143104779 Chr.X: 154700023-154700023 on Build GRCh38
rs371718474 Chr.X: 154700015-154700015 on Build GRCh38
rs376742477 Chr.X: 154700084-154700084 on Build GRCh38
rs202124648 Chr.X: 154700086-154700086 on Build GRCh38
rs369725320 Chr.X: 154700094-154700094 on Build GRCh38
rs782273112 Chr.X: 154700137-154700137 on Build GRCh38
rs374145424 Chr.X: 154700105-154700105 on Build GRCh38
rs782328386 Chr.X: 154700135-154700135 on Build GRCh38
rs782122393 Chr.X: 154699985-154699985 on Build GRCh38
rs782580659 Chr.X: 154700038-154700038 on Build GRCh38
rs6643691 Chr.X: 154700119-154700119 on Build GRCh38
rs782360417 Chr.X: 154700010-154700010 on Build GRCh38
rs781837912 Chr.X: 154699941-154699941 on Build GRCh38
rs782485246 Chr.X: 154700096-154700096 on Build GRCh38
rs781895883 Chr.X: 154700057-154700057 on Build GRCh38
rs782721213 Chr.X: 154700102-154700102 on Build GRCh38
rs7888328 Chr.X: 154699944-154699944 on Build GRCh38
rs896192586 Chr.X: 154700070-154700070 on Build GRCh38
rs374878750 Chr.X: 154699970-154699970 on Build GRCh38
rs782054509 Chr.X: 154700025-154700025 on Build GRCh38

View Full Product Details