Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40737289-40737520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222010 c.1264G>A p.V422I Chr.15: 40737302-40737302 on Build GRCh38
7212264 c.1225-3C>G p.? Chr.15: 40737344-40737344 on Build GRCh38
244404 c.1268A>G p.Y423C Chr.15: 40737298-40737298 on Build GRCh38
7317886 c.1235T>A p.L412Q Chr.15: 40737331-40737331 on Build GRCh38
5549810 c.1257A>C p.A419A Chr.15: 40737309-40737309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773916777 Chr.15: 40737318-40737318 on Build GRCh38
rs2304581 Chr.15: 40737480-40737480 on Build GRCh38
rs912280200 Chr.15: 40737388-40737388 on Build GRCh38
rs116351777 Chr.15: 40737381-40737381 on Build GRCh38
rs1022866178 Chr.15: 40737487-40737487 on Build GRCh38
rs903502796 Chr.15: 40737444-40737444 on Build GRCh38
rs2304580 Chr.15: 40737469-40737469 on Build GRCh38
rs936344877 Chr.15: 40737447-40737447 on Build GRCh38
rs59558714 Chr.15: 40737494-40737494 on Build GRCh38
rs1056476744 Chr.15: 40737463-40737463 on Build GRCh38
rs753674994 Chr.15: 40737378-40737378 on Build GRCh38
rs112493418 Chr.15: 40737385-40737385 on Build GRCh38
rs778257020 Chr.15: 40737355-40737356 on Build GRCh38
rs763833521 Chr.15: 40737374-40737374 on Build GRCh38
rs199947975 Chr.15: 40737359-40737359 on Build GRCh38
rs561800012 Chr.15: 40737325-40737325 on Build GRCh38
rs1039342937 Chr.15: 40737405-40737405 on Build GRCh38
rs200189558 Chr.15: 40737341-40737341 on Build GRCh38
rs772056920 Chr.15: 40737332-40737332 on Build GRCh38
rs775239003 Chr.15: 40737344-40737344 on Build GRCh38
rs749733702 Chr.15: 40737387-40737388 on Build GRCh38
rs920881094 Chr.15: 40737357-40737357 on Build GRCh38
rs140424662 Chr.15: 40737399-40737399 on Build GRCh38

View Full Product Details