Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55870837-55871082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
UniGene
Hs.269665
Gene Chromosome Location
Chr.4: 55853616-55905078 on Build GRCh38
Transcripts
NM_001024924.1 NM_018261.3 NM_178237.2 XM_005265747.2 XM_005265748.1 XM_005265750.2 XM_017008405.1 XM_017008406.1 XM_017008407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731804 c.765C>A p.S255R Chr.4: 55870839-55870839 on Build GRCh38
4681998 c.781C>A p.L261I Chr.4: 55870855-55870855 on Build GRCh38
1284791 c.816G>A p.E272E Chr.4: 55870890-55870890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766247298 Chr.4: 55870870-55870870 on Build GRCh38
rs762433095 Chr.4: 55870903-55870903 on Build GRCh38
rs755975266 Chr.4: 55870914-55870914 on Build GRCh38
rs951800849 Chr.4: 55870961-55870961 on Build GRCh38
rs756482120 Chr.4: 55870927-55870927 on Build GRCh38
rs937720360 Chr.4: 55870985-55870985 on Build GRCh38
rs200303000 Chr.4: 55870930-55870931 on Build GRCh38
rs906272964 Chr.4: 55870945-55870945 on Build GRCh38
rs1055191755 Chr.4: 55871003-55871003 on Build GRCh38
rs751278702 Chr.4: 55870930-55870930 on Build GRCh38
rs557931911 Chr.4: 55870931-55870931 on Build GRCh38
rs369619617 Chr.4: 55870949-55870949 on Build GRCh38
rs1007753387 Chr.4: 55871005-55871005 on Build GRCh38
rs200395524 Chr.4: 55870884-55870884 on Build GRCh38
rs372578105 Chr.4: 55870917-55870917 on Build GRCh38
rs759039846 Chr.4: 55870910-55870910 on Build GRCh38
rs752377692 Chr.4: 55870898-55870898 on Build GRCh38
rs763662120 Chr.4: 55870909-55870909 on Build GRCh38
rs751053841 Chr.4: 55870911-55870911 on Build GRCh38
rs764845637 Chr.4: 55870893-55870893 on Build GRCh38
rs899452936 Chr.4: 55871036-55871036 on Build GRCh38
rs375609735 Chr.4: 55870918-55870918 on Build GRCh38

View Full Product Details