Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7690283-7690540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445982 c.621+3G>A p.? Chr.19: 7690403-7690403 on Build GRCh38
6580999 c.574G>A p.D192N Chr.19: 7690453-7690453 on Build GRCh38
1751266 c.521G>C p.C174S Chr.19: 7690506-7690506 on Build GRCh38
3766462 c.621+7T>C p.? Chr.19: 7690399-7690399 on Build GRCh38
4581581 c.563G>A p.R188Q Chr.19: 7690464-7690464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751787016 Chr.19: 7690506-7690506 on Build GRCh38
rs748703601 Chr.19: 7690450-7690450 on Build GRCh38
rs1038551504 Chr.19: 7690440-7690440 on Build GRCh38
rs757378683 Chr.19: 7690316-7690316 on Build GRCh38
rs770755660 Chr.19: 7690468-7690468 on Build GRCh38
rs746104507 Chr.19: 7690357-7690357 on Build GRCh38
rs769204272 Chr.19: 7690370-7690370 on Build GRCh38
rs753914101 Chr.19: 7690311-7690311 on Build GRCh38
rs768462140 Chr.19: 7690500-7690500 on Build GRCh38
rs139487727 Chr.19: 7690475-7690475 on Build GRCh38
rs182569239 Chr.19: 7690497-7690497 on Build GRCh38
rs957194543 Chr.19: 7690395-7690395 on Build GRCh38
rs542938135 Chr.19: 7690351-7690351 on Build GRCh38
rs148383467 Chr.19: 7690355-7690356 on Build GRCh38
rs764237783 Chr.19: 7690309-7690309 on Build GRCh38
rs911429952 Chr.19: 7690495-7690495 on Build GRCh38
rs989938926 Chr.19: 7690397-7690397 on Build GRCh38
rs781488649 Chr.19: 7690447-7690447 on Build GRCh38
rs767633435 Chr.19: 7690402-7690403 on Build GRCh38
rs565338813 Chr.19: 7690416-7690416 on Build GRCh38
rs144440869 Chr.19: 7690451-7690451 on Build GRCh38
rs28364072 Chr.19: 7690399-7690399 on Build GRCh38
rs200648991 Chr.19: 7690464-7690464 on Build GRCh38
rs377564979 Chr.19: 7690411-7690411 on Build GRCh38
rs758701728 Chr.19: 7690361-7690361 on Build GRCh38
rs750593910 Chr.19: 7690421-7690421 on Build GRCh38
rs771142988 Chr.19: 7690432-7690432 on Build GRCh38
rs755369130 Chr.19: 7690446-7690446 on Build GRCh38
rs959158304 Chr.19: 7690304-7690304 on Build GRCh38
rs990622247 Chr.19: 7690312-7690312 on Build GRCh38
rs748919449 Chr.19: 7690465-7690465 on Build GRCh38
rs149647877 Chr.19: 7690348-7690348 on Build GRCh38
rs774327816 Chr.19: 7690404-7690404 on Build GRCh38
rs771903938 Chr.19: 7690484-7690484 on Build GRCh38
rs763047133 Chr.19: 7690499-7690499 on Build GRCh38
rs373313846 Chr.19: 7690379-7690379 on Build GRCh38
rs267605763 Chr.19: 7690483-7690483 on Build GRCh38
rs774169912 Chr.19: 7690472-7690472 on Build GRCh38
rs867413961 Chr.19: 7690444-7690444 on Build GRCh38
rs772683395 Chr.19: 7690398-7690398 on Build GRCh38
rs138319893 Chr.19: 7690308-7690308 on Build GRCh38
rs4996972 Chr.19: 7690327-7690327 on Build GRCh38
rs778961760 Chr.19: 7690356-7690356 on Build GRCh38
rs773302708 Chr.19: 7690361-7690362 on Build GRCh38
rs369443024 Chr.19: 7690417-7690417 on Build GRCh38
rs375077946 Chr.19: 7690415-7690415 on Build GRCh38
rs377575240 Chr.19: 7690502-7690502 on Build GRCh38
rs375114070 Chr.19: 7690367-7690367 on Build GRCh38
rs758952802 Chr.19: 7690441-7690441 on Build GRCh38
rs145289159 Chr.19: 7690513-7690513 on Build GRCh38
rs528496860 Chr.19: 7690368-7690368 on Build GRCh38
rs141227304 Chr.19: 7690405-7690405 on Build GRCh38
rs191142459 Chr.19: 7690372-7690372 on Build GRCh38
rs536515427 Chr.19: 7690496-7690496 on Build GRCh38
rs368929610 Chr.19: 7690512-7690512 on Build GRCh38
rs770845770 Chr.19: 7690403-7690403 on Build GRCh38
rs72558010 Chr.19: 7690328-7690328 on Build GRCh38
rs934015813 Chr.19: 7690373-7690373 on Build GRCh38
rs758683190 Chr.19: 7690428-7690428 on Build GRCh38
rs373413003 Chr.19: 7690494-7690494 on Build GRCh38
rs751530180 Chr.19: 7690305-7690305 on Build GRCh38
rs765369748 Chr.19: 7690418-7690418 on Build GRCh38

View Full Product Details