Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38496331-38496584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOG
Entrez Gene ID
9941
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB
UniGene
Hs.517897
Gene Chromosome Location
Chr.3: 38496127-38526305 on Build GRCh38
Transcripts
NM_001145464.1 NM_001318954.1 NM_001318955.1 NM_001318956.1 NM_001318957.1 NM_001318958.1 NM_001318959.1 NM_005107.3 NR_134938.1 XM_005265633.3 XM_005265635.3 XM_006713442.3 XM_006713444.3 XM_017007584.1 XM_017007585.1 XM_017007586.1 XM_017007587.1 XM_017007588.1 XM_017007589.1 XM_017007590.1 XM_017007591.1 XM_017007592.1 XM_017007593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7168845 c.18C>T p.I6I Chr.3: 38496385-38496385 on Build GRCh38
1745170 c.130_138delGCTGAGGGA p.E45_A47delEGA Chr.3: 38496497-38496505 on Build GRCh38
6164846 c.74T>C p.V25A Chr.3: 38496441-38496441 on Build GRCh38
7242732 c.60C>T p.F20F Chr.3: 38496427-38496427 on Build GRCh38
2152547 c.71_72delCT p.A24fs*21 Chr.3: 38496438-38496439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779339742 Chr.3: 38496506-38496506 on Build GRCh38
rs756161009 Chr.3: 38496376-38496376 on Build GRCh38
rs752387567 Chr.3: 38496474-38496474 on Build GRCh38
rs773707653 Chr.3: 38496364-38496364 on Build GRCh38
rs767309530 Chr.3: 38496530-38496530 on Build GRCh38
rs956084161 Chr.3: 38496520-38496520 on Build GRCh38
rs370106287 Chr.3: 38496464-38496464 on Build GRCh38
rs775712685 Chr.3: 38496517-38496517 on Build GRCh38
rs766432267 Chr.3: 38496525-38496525 on Build GRCh38
rs758855102 Chr.3: 38496449-38496457 on Build GRCh38
rs750383065 Chr.3: 38496379-38496379 on Build GRCh38
rs373909153 Chr.3: 38496559-38496559 on Build GRCh38
rs760734069 Chr.3: 38496431-38496431 on Build GRCh38
rs938305955 Chr.3: 38496402-38496402 on Build GRCh38
rs765146335 Chr.3: 38496377-38496377 on Build GRCh38
rs779464079 Chr.3: 38496554-38496554 on Build GRCh38
rs368855859 Chr.3: 38496422-38496422 on Build GRCh38
rs371730440 Chr.3: 38496395-38496395 on Build GRCh38
rs200468418 Chr.3: 38496427-38496427 on Build GRCh38
rs751524406 Chr.3: 38496527-38496527 on Build GRCh38
rs779647446 Chr.3: 38496544-38496545 on Build GRCh38
rs755617306 Chr.3: 38496486-38496486 on Build GRCh38
rs1025061181 Chr.3: 38496378-38496378 on Build GRCh38
rs142607341 Chr.3: 38496405-38496405 on Build GRCh38
rs751348639 Chr.3: 38496392-38496392 on Build GRCh38
rs750909137 Chr.3: 38496412-38496412 on Build GRCh38
rs763363128 Chr.3: 38496366-38496366 on Build GRCh38
rs149413458 Chr.3: 38496443-38496443 on Build GRCh38
rs750432385 Chr.3: 38496439-38496439 on Build GRCh38
rs899061416 Chr.3: 38496472-38496472 on Build GRCh38
rs770202760 Chr.3: 38496363-38496363 on Build GRCh38
rs761916512 Chr.3: 38496523-38496523 on Build GRCh38
rs377680068 Chr.3: 38496521-38496521 on Build GRCh38
rs138226578 Chr.3: 38496426-38496426 on Build GRCh38
rs762160473 Chr.3: 38496377-38496378 on Build GRCh38
rs747654799 Chr.3: 38496361-38496361 on Build GRCh38
rs372608773 Chr.3: 38496547-38496547 on Build GRCh38
rs989147841 Chr.3: 38496407-38496407 on Build GRCh38
rs368861888 Chr.3: 38496373-38496373 on Build GRCh38
rs778315577 Chr.3: 38496398-38496398 on Build GRCh38
rs746276326 Chr.3: 38496507-38496507 on Build GRCh38
rs771436629 Chr.3: 38496499-38496499 on Build GRCh38
rs775615944 Chr.3: 38496510-38496510 on Build GRCh38
rs766378816 Chr.3: 38496441-38496441 on Build GRCh38
rs138441261 Chr.3: 38496466-38496466 on Build GRCh38
rs764205454 Chr.3: 38496434-38496434 on Build GRCh38
rs746626484 Chr.3: 38496350-38496350 on Build GRCh38
rs765771875 Chr.3: 38496387-38496388 on Build GRCh38
rs1032877972 Chr.3: 38496352-38496352 on Build GRCh38
rs184615397 Chr.3: 38496565-38496565 on Build GRCh38
rs201317819 Chr.3: 38496387-38496387 on Build GRCh38
rs373135233 Chr.3: 38496513-38496513 on Build GRCh38
rs116479475 Chr.3: 38496544-38496544 on Build GRCh38
rs772481733 Chr.3: 38496509-38496509 on Build GRCh38
rs753363770 Chr.3: 38496550-38496550 on Build GRCh38
rs767742733 Chr.3: 38496375-38496375 on Build GRCh38
rs916982789 Chr.3: 38496374-38496374 on Build GRCh38
rs748300582 Chr.3: 38496368-38496368 on Build GRCh38
rs763005298 Chr.3: 38496440-38496440 on Build GRCh38
rs140624245 Chr.3: 38496410-38496410 on Build GRCh38
rs754739929 Chr.3: 38496461-38496461 on Build GRCh38
rs34327004 Chr.3: 38496502-38496502 on Build GRCh38
rs766723778 Chr.3: 38496371-38496371 on Build GRCh38
rs139206818 Chr.3: 38496514-38496514 on Build GRCh38
rs199919669 Chr.3: 38496354-38496354 on Build GRCh38
rs571342831 Chr.3: 38496562-38496562 on Build GRCh38
rs780830129 Chr.3: 38496393-38496393 on Build GRCh38
rs756746296 Chr.3: 38496552-38496552 on Build GRCh38
rs193285202 Chr.3: 38496546-38496546 on Build GRCh38
rs748759280 Chr.3: 38496495-38496495 on Build GRCh38
rs138933023 Chr.3: 38496437-38496437 on Build GRCh38
rs779921105 Chr.3: 38496348-38496348 on Build GRCh38
rs759312453 Chr.3: 38496528-38496528 on Build GRCh38
rs759832007 Chr.3: 38496421-38496421 on Build GRCh38
rs371857709 Chr.3: 38496383-38496383 on Build GRCh38
rs202033501 Chr.3: 38496357-38496357 on Build GRCh38
rs746356754 Chr.3: 38496557-38496557 on Build GRCh38
rs754076258 Chr.3: 38496473-38496473 on Build GRCh38
rs955885366 Chr.3: 38496359-38496359 on Build GRCh38
rs777288999 Chr.3: 38496489-38496489 on Build GRCh38
rs145619725 Chr.3: 38496394-38496394 on Build GRCh38

View Full Product Details