Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4310238-4310511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1
Entrez Gene ID
79187
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
GLFND, MIR1
UniGene
Hs.28144
Gene Chromosome Location
Chr.19: 4304594-4323846 on Build GRCh38
Transcripts
NM_024333.2 XM_005259644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161162 c.402G>C p.L134F Chr.19: 4310508-4310508 on Build GRCh38
4398354 c.404A>G p.K135R Chr.19: 4310510-4310510 on Build GRCh38
6865213 c.362A>C p.K121T Chr.19: 4310289-4310289 on Build GRCh38
329000 c.367G>A p.G123R Chr.19: 4310294-4310294 on Build GRCh38
6385730 c.403A>G p.K135E Chr.19: 4310509-4310509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775060072 Chr.19: 4310477-4310482 on Build GRCh38
rs1040962633 Chr.19: 4310411-4310411 on Build GRCh38
rs537282275 Chr.19: 4310396-4310396 on Build GRCh38
rs376561346 Chr.19: 4310454-4310454 on Build GRCh38
rs753687871 Chr.19: 4310337-4310337 on Build GRCh38
rs60165433 Chr.19: 4310359-4310360 on Build GRCh38
rs11879976 Chr.19: 4310330-4310330 on Build GRCh38
rs377023321 Chr.19: 4310356-4310356 on Build GRCh38
rs557099158 Chr.19: 4310417-4310417 on Build GRCh38
rs370384771 Chr.19: 4310448-4310448 on Build GRCh38
rs957616628 Chr.19: 4310423-4310423 on Build GRCh38
rs754684683 Chr.19: 4310444-4310444 on Build GRCh38
rs912582976 Chr.19: 4310347-4310347 on Build GRCh38
rs569675562 Chr.19: 4310418-4310418 on Build GRCh38
rs555080190 Chr.19: 4310430-4310430 on Build GRCh38
rs774618825 Chr.19: 4310324-4310324 on Build GRCh38
rs1038264264 Chr.19: 4310298-4310298 on Build GRCh38
rs758048713 Chr.19: 4310429-4310429 on Build GRCh38
rs551706022 Chr.19: 4310380-4310380 on Build GRCh38
rs549130691 Chr.19: 4310306-4310306 on Build GRCh38
rs372218316 Chr.19: 4310490-4310490 on Build GRCh38
rs200369660 Chr.19: 4310478-4310478 on Build GRCh38
rs760486782 Chr.19: 4310326-4310326 on Build GRCh38
rs199652161 Chr.19: 4310457-4310457 on Build GRCh38
rs746712009 Chr.19: 4310434-4310434 on Build GRCh38
rs766570708 Chr.19: 4310308-4310308 on Build GRCh38
rs771659397 Chr.19: 4310488-4310488 on Build GRCh38
rs76786296 Chr.19: 4310424-4310424 on Build GRCh38
rs1049068819 Chr.19: 4310410-4310410 on Build GRCh38
rs200582682 Chr.19: 4310303-4310303 on Build GRCh38
rs1019672756 Chr.19: 4310487-4310487 on Build GRCh38
rs757101077 Chr.19: 4310340-4310340 on Build GRCh38
rs921556013 Chr.19: 4310392-4310392 on Build GRCh38
rs550870777 Chr.19: 4310358-4310358 on Build GRCh38
rs375127336 Chr.19: 4310453-4310453 on Build GRCh38
rs367825820 Chr.19: 4310364-4310364 on Build GRCh38
rs767810155 Chr.19: 4310482-4310482 on Build GRCh38
rs770366923 Chr.19: 4310279-4310279 on Build GRCh38
rs377370376 Chr.19: 4310447-4310447 on Build GRCh38
rs1033248514 Chr.19: 4310419-4310419 on Build GRCh38
rs931575617 Chr.19: 4310397-4310397 on Build GRCh38
rs774533083 Chr.19: 4310459-4310459 on Build GRCh38
rs528790211 Chr.19: 4310342-4310342 on Build GRCh38
rs759627069 Chr.19: 4310463-4310463 on Build GRCh38
rs145281893 Chr.19: 4310485-4310485 on Build GRCh38
rs571586888 Chr.19: 4310391-4310391 on Build GRCh38
rs371390445 Chr.19: 4310486-4310486 on Build GRCh38
rs776233464 Chr.19: 4310483-4310483 on Build GRCh38
rs1007939428 Chr.19: 4310375-4310375 on Build GRCh38
rs957032809 Chr.19: 4310274-4310274 on Build GRCh38

View Full Product Details