Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94496279-94496538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305418 c.4318C>T p.Q1440* Chr.8: 94496393-94496393 on Build GRCh38
4616055 c.4254_4257delTCTC p.L1419fs*3 Chr.8: 94496454-94496457 on Build GRCh38
4616056 c.4250_4251insG p.N1417fs*18 Chr.8: 94496460-94496461 on Build GRCh38
1102583 c.4336G>T p.E1446* Chr.8: 94496375-94496375 on Build GRCh38
4656619 c.4252G>A p.G1418S Chr.8: 94496459-94496459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371099063 Chr.8: 94496443-94496443 on Build GRCh38
rs771810853 Chr.8: 94496427-94496427 on Build GRCh38
rs766722548 Chr.8: 94496371-94496371 on Build GRCh38
rs776192620 Chr.8: 94496307-94496307 on Build GRCh38
rs2289455 Chr.8: 94496493-94496493 on Build GRCh38
rs560304943 Chr.8: 94496475-94496475 on Build GRCh38
rs745666981 Chr.8: 94496425-94496425 on Build GRCh38
rs757658819 Chr.8: 94496312-94496312 on Build GRCh38
rs751963634 Chr.8: 94496374-94496374 on Build GRCh38
rs745306929 Chr.8: 94496500-94496500 on Build GRCh38
rs533039697 Chr.8: 94496428-94496428 on Build GRCh38
rs139309146 Chr.8: 94496454-94496454 on Build GRCh38
rs369048370 Chr.8: 94496306-94496306 on Build GRCh38
rs78686282 Chr.8: 94496387-94496387 on Build GRCh38
rs776203727 Chr.8: 94496442-94496442 on Build GRCh38
rs374090382 Chr.8: 94496446-94496446 on Build GRCh38
rs768095186 Chr.8: 94496436-94496436 on Build GRCh38
rs35530463 Chr.8: 94496350-94496351 on Build GRCh38
rs368083067 Chr.8: 94496464-94496464 on Build GRCh38
rs868444716 Chr.8: 94496431-94496431 on Build GRCh38
rs1017976081 Chr.8: 94496364-94496364 on Build GRCh38
rs765471362 Chr.8: 94496504-94496504 on Build GRCh38
rs750590650 Chr.8: 94496507-94496507 on Build GRCh38
rs757486870 Chr.8: 94496421-94496421 on Build GRCh38
rs1007039178 Chr.8: 94496343-94496343 on Build GRCh38
rs777887809 Chr.8: 94496405-94496405 on Build GRCh38
rs545261875 Chr.8: 94496339-94496339 on Build GRCh38
rs753216437 Chr.8: 94496391-94496391 on Build GRCh38
rs183217637 Chr.8: 94496430-94496430 on Build GRCh38
rs920853917 Chr.8: 94496414-94496414 on Build GRCh38
rs778945233 Chr.8: 94496422-94496422 on Build GRCh38
rs754427802 Chr.8: 94496409-94496409 on Build GRCh38
rs569055401 Chr.8: 94496498-94496498 on Build GRCh38
rs576719748 Chr.8: 94496338-94496338 on Build GRCh38
rs79992272 Chr.8: 94496305-94496305 on Build GRCh38
rs781768757 Chr.8: 94496320-94496320 on Build GRCh38
rs150870867 Chr.8: 94496397-94496397 on Build GRCh38
rs757215885 Chr.8: 94496503-94496503 on Build GRCh38
rs759925259 Chr.8: 94496384-94496384 on Build GRCh38
rs371279436 Chr.8: 94496408-94496408 on Build GRCh38
rs374750145 Chr.8: 94496496-94496496 on Build GRCh38
rs776189137 Chr.8: 94496473-94496473 on Build GRCh38
rs200591653 Chr.8: 94496508-94496508 on Build GRCh38

View Full Product Details