Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4506844-4507115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMOX2
Entrez Gene ID
3163
Gene Name
heme oxygenase 2
Gene Aliases
HO-2
UniGene
Hs.284279
Gene Chromosome Location
Chr.16: 4474697-4510347 on Build GRCh38
Transcripts
NM_001127204.1 NM_001127205.1 NM_001127206.2 NM_001286267.1 NM_001286268.1 NM_001286269.1 NM_001286270.1 NM_001286271.1 NM_002134.3 XM_011522473.1 XM_017023196.1 XM_017023197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681118 c.140G>A p.R47Q Chr.16: 4506948-4506948 on Build GRCh38
7218611 c.87-10C>G p.? Chr.16: 4506885-4506885 on Build GRCh38
6681119 c.90G>T p.M30I Chr.16: 4506898-4506898 on Build GRCh38
970717 c.114G>T p.K38N Chr.16: 4506922-4506922 on Build GRCh38
1301970 c.87-1G>A p.? Chr.16: 4506894-4506894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149978208 Chr.16: 4506964-4506964 on Build GRCh38
rs777612788 Chr.16: 4506905-4506905 on Build GRCh38
rs879236886 Chr.16: 4507063-4507063 on Build GRCh38
rs776861637 Chr.16: 4506950-4506950 on Build GRCh38
rs889257204 Chr.16: 4507090-4507090 on Build GRCh38
rs150902900 Chr.16: 4506948-4506948 on Build GRCh38
rs768922150 Chr.16: 4506882-4506882 on Build GRCh38
rs761920849 Chr.16: 4506884-4506884 on Build GRCh38
rs780898232 Chr.16: 4507027-4507027 on Build GRCh38
rs372362103 Chr.16: 4506903-4506903 on Build GRCh38
rs374647263 Chr.16: 4507023-4507023 on Build GRCh38
rs917711821 Chr.16: 4507050-4507050 on Build GRCh38
rs760898988 Chr.16: 4506868-4506868 on Build GRCh38
rs11542539 Chr.16: 4506959-4506959 on Build GRCh38
rs144293425 Chr.16: 4506998-4506998 on Build GRCh38
rs764664140 Chr.16: 4507011-4507011 on Build GRCh38
rs753522599 Chr.16: 4506909-4506909 on Build GRCh38
rs752086119 Chr.16: 4506990-4506990 on Build GRCh38
rs544701346 Chr.16: 4506883-4506883 on Build GRCh38
rs960107117 Chr.16: 4507035-4507035 on Build GRCh38
rs1046794064 Chr.16: 4507020-4507020 on Build GRCh38
rs757840609 Chr.16: 4507014-4507014 on Build GRCh38
rs200587307 Chr.16: 4507024-4507024 on Build GRCh38
rs139279048 Chr.16: 4506958-4506958 on Build GRCh38
rs745794435 Chr.16: 4506943-4506943 on Build GRCh38
rs991993906 Chr.16: 4507047-4507047 on Build GRCh38
rs753571806 Chr.16: 4507001-4507001 on Build GRCh38
rs778442977 Chr.16: 4506941-4506941 on Build GRCh38
rs576788137 Chr.16: 4506889-4506889 on Build GRCh38
rs752031128 Chr.16: 4506899-4506899 on Build GRCh38
rs878912853 Chr.16: 4507062-4507062 on Build GRCh38
rs1037851391 Chr.16: 4507031-4507031 on Build GRCh38
rs11542537 Chr.16: 4506961-4506961 on Build GRCh38
rs779840322 Chr.16: 4506947-4506947 on Build GRCh38
rs368272506 Chr.16: 4506886-4506886 on Build GRCh38
rs773286905 Chr.16: 4506960-4506960 on Build GRCh38
rs533385221 Chr.16: 4506926-4506926 on Build GRCh38
rs200960043 Chr.16: 4506944-4506944 on Build GRCh38
rs767940988 Chr.16: 4507000-4507000 on Build GRCh38
rs566853006 Chr.16: 4507087-4507087 on Build GRCh38
rs898123891 Chr.16: 4506892-4506892 on Build GRCh38
rs766667349 Chr.16: 4506890-4506890 on Build GRCh38
rs375791662 Chr.16: 4507056-4507056 on Build GRCh38
rs749428526 Chr.16: 4507057-4507057 on Build GRCh38
rs25685 Chr.16: 4506910-4506910 on Build GRCh38
rs748334381 Chr.16: 4507046-4507046 on Build GRCh38
rs140785119 Chr.16: 4507002-4507002 on Build GRCh38
rs770113488 Chr.16: 4507051-4507051 on Build GRCh38
rs754799686 Chr.16: 4507025-4507025 on Build GRCh38
rs947972787 Chr.16: 4506873-4506873 on Build GRCh38
rs981068956 Chr.16: 4507089-4507089 on Build GRCh38

View Full Product Details