Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127595935-127596162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN3
Entrez Gene ID
29999
Gene Name
fascin actin-bundling protein 3
Gene Aliases
-
UniGene
Hs.712692
Gene Chromosome Location
Chr.7: 127593635-127601797 on Build GRCh38
Transcripts
NM_020369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5581139 c.841+1G>A p.? Chr.7: 127596004-127596004 on Build GRCh38
1673515 c.820C>T p.R274W Chr.7: 127595982-127595982 on Build GRCh38
7245312 c.821G>T p.R274L Chr.7: 127595983-127595983 on Build GRCh38
6548715 c.782A>T p.H261L Chr.7: 127595944-127595944 on Build GRCh38
357071 c.817G>T p.G273W Chr.7: 127595979-127595979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770327743 Chr.7: 127596001-127596001 on Build GRCh38
rs752537878 Chr.7: 127595970-127595970 on Build GRCh38
rs754535550 Chr.7: 127596130-127596130 on Build GRCh38
rs956203911 Chr.7: 127596136-127596136 on Build GRCh38
rs781357092 Chr.7: 127595992-127595992 on Build GRCh38
rs550557390 Chr.7: 127596063-127596063 on Build GRCh38
rs917272763 Chr.7: 127596117-127596117 on Build GRCh38
rs753360467 Chr.7: 127596027-127596027 on Build GRCh38
rs543035516 Chr.7: 127596054-127596054 on Build GRCh38
rs769012355 Chr.7: 127596017-127596017 on Build GRCh38
rs745971352 Chr.7: 127595997-127595997 on Build GRCh38
rs765995498 Chr.7: 127596025-127596025 on Build GRCh38
rs992710852 Chr.7: 127596095-127596095 on Build GRCh38
rs758326784 Chr.7: 127595971-127595971 on Build GRCh38
rs904556177 Chr.7: 127596014-127596014 on Build GRCh38
rs772811300 Chr.7: 127596018-127596018 on Build GRCh38
rs765208316 Chr.7: 127596031-127596031 on Build GRCh38
rs760295617 Chr.7: 127596113-127596114 on Build GRCh38
rs747243388 Chr.7: 127595982-127595982 on Build GRCh38
rs765897657 Chr.7: 127596024-127596027 on Build GRCh38
rs753668205 Chr.7: 127595955-127595955 on Build GRCh38
rs758205809 Chr.7: 127596050-127596050 on Build GRCh38
rs73720862 Chr.7: 127595962-127595962 on Build GRCh38
rs759036110 Chr.7: 127596029-127596029 on Build GRCh38
rs778757556 Chr.7: 127595968-127595968 on Build GRCh38
rs376914564 Chr.7: 127596003-127596003 on Build GRCh38
rs569716722 Chr.7: 127595983-127595983 on Build GRCh38
rs200608418 Chr.7: 127596026-127596026 on Build GRCh38
rs752695466 Chr.7: 127596048-127596048 on Build GRCh38
rs1000390954 Chr.7: 127596090-127596090 on Build GRCh38
rs760396638 Chr.7: 127596022-127596022 on Build GRCh38
rs778083885 Chr.7: 127595972-127595972 on Build GRCh38

View Full Product Details